Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 106823213-106823440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D8B
Entrez Gene ID
54885
Gene Name
TBC1 domain family member 8B
Gene Aliases
GRAMD8B
UniGene
Hs.351798
Gene Chromosome Location
Chr.X: 106802689-106876150 on Build GRCh38
Transcripts
NM_017752.2 NM_198881.1 XM_006724669.1 XM_006724670.3

SNP Detail

SNP ID Chromosome Location
rs772217563 Chr.X: 106823311-106823311 on Build GRCh38
rs749148459 Chr.X: 106823370-106823370 on Build GRCh38
rs769456918 Chr.X: 106823356-106823356 on Build GRCh38
rs761410195 Chr.X: 106823377-106823377 on Build GRCh38
rs772601210 Chr.X: 106823367-106823367 on Build GRCh38
rs186913793 Chr.X: 106823302-106823302 on Build GRCh38
rs750065146 Chr.X: 106823386-106823386 on Build GRCh38
rs1008740907 Chr.X: 106823348-106823348 on Build GRCh38
rs767219068 Chr.X: 106823320-106823320 on Build GRCh38
rs748241860 Chr.X: 106823303-106823303 on Build GRCh38
rs766157549 Chr.X: 106823244-106823244 on Build GRCh38
rs756083514 Chr.X: 106823270-106823270 on Build GRCh38
rs755546236 Chr.X: 106823298-106823298 on Build GRCh38
rs760425057 Chr.X: 106823412-106823412 on Build GRCh38
rs140106583 Chr.X: 106823381-106823381 on Build GRCh38
rs749735768 Chr.X: 106823256-106823256 on Build GRCh38
rs780337633 Chr.X: 106823400-106823400 on Build GRCh38
rs759889636 Chr.X: 106823290-106823291 on Build GRCh38
rs745629515 Chr.X: 106823353-106823353 on Build GRCh38
rs773339487 Chr.X: 106823313-106823313 on Build GRCh38
rs775398836 Chr.X: 106823365-106823365 on Build GRCh38

View Full Product Details