Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 19708105-19708336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM5D
Entrez Gene ID
8284
Gene Name
lysine demethylase 5D
Gene Aliases
HY, HYA, JARID1D, SMCY
UniGene
Hs.80358
Gene Chromosome Location
Chr.Y: 19692495-19745347 on Build GRCh38
Transcripts
NM_001146705.1 NM_001146706.1 NM_004653.4 XM_005262560.2 XM_005262561.2 XM_011531468.2

SNP Detail

SNP ID Chromosome Location
rs748855039 Chr.Y: 19708291-19708291 on Build GRCh38
rs924216272 Chr.Y: 19708131-19708131 on Build GRCh38
rs757563188 Chr.Y: 19708181-19708181 on Build GRCh38
rs746183701 Chr.Y: 19708236-19708236 on Build GRCh38
rs752105810 Chr.Y: 19708195-19708195 on Build GRCh38
rs757605600 Chr.Y: 19708229-19708229 on Build GRCh38
rs755795687 Chr.Y: 19708249-19708249 on Build GRCh38
rs781516859 Chr.Y: 19708230-19708230 on Build GRCh38
rs779615383 Chr.Y: 19708272-19708272 on Build GRCh38

View Full Product Details