Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93186499-93186724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM251
Entrez Gene ID
26175
Gene Name
transmembrane protein 251
Gene Aliases
C14orf109
UniGene
Hs.275352
Gene Chromosome Location
Chr.14: 93184951-93187086 on Build GRCh38
Transcripts
NM_001098621.1 NM_015676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370102 c.284A>G p.K95R Chr.14: 93186559-93186559 on Build GRCh38
3499052 c.290T>C p.V97A Chr.14: 93186565-93186565 on Build GRCh38
1587699 c.490C>G p.L164V Chr.14: 93186651-93186651 on Build GRCh38
3499054 c.383T>C p.L128P Chr.14: 93186658-93186658 on Build GRCh38
3499050 c.230T>A p.V77E Chr.14: 93186505-93186505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979650011 Chr.14: 93186631-93186631 on Build GRCh38
rs897996630 Chr.14: 93186539-93186539 on Build GRCh38
rs764827345 Chr.14: 93186550-93186550 on Build GRCh38
rs750710970 Chr.14: 93186594-93186594 on Build GRCh38
rs374164576 Chr.14: 93186667-93186667 on Build GRCh38
rs774992621 Chr.14: 93186690-93186690 on Build GRCh38
rs372120375 Chr.14: 93186647-93186647 on Build GRCh38
rs769224822 Chr.14: 93186685-93186685 on Build GRCh38
rs1017594036 Chr.14: 93186601-93186601 on Build GRCh38
rs202152856 Chr.14: 93186604-93186604 on Build GRCh38
rs887886820 Chr.14: 93186590-93186590 on Build GRCh38
rs745564025 Chr.14: 93186679-93186679 on Build GRCh38
rs535664901 Chr.14: 93186668-93186668 on Build GRCh38
rs780314112 Chr.14: 93186607-93186607 on Build GRCh38
rs756102043 Chr.14: 93186600-93186600 on Build GRCh38
rs765919628 Chr.14: 93186583-93186583 on Build GRCh38
rs901849912 Chr.14: 93186606-93186606 on Build GRCh38
rs367611847 Chr.14: 93186540-93186540 on Build GRCh38
rs775937774 Chr.14: 93186555-93186555 on Build GRCh38
rs115343271 Chr.14: 93186585-93186585 on Build GRCh38
rs374665855 Chr.14: 93186591-93186591 on Build GRCh38
rs764576645 Chr.14: 93186696-93186696 on Build GRCh38
rs1032157064 Chr.14: 93186642-93186642 on Build GRCh38
rs748034329 Chr.14: 93186610-93186610 on Build GRCh38
rs145078947 Chr.14: 93186629-93186629 on Build GRCh38
rs751911577 Chr.14: 93186584-93186584 on Build GRCh38
rs929429999 Chr.14: 93186552-93186552 on Build GRCh38
rs539005733 Chr.14: 93186581-93186581 on Build GRCh38
rs747199383 Chr.14: 93186604-93186604 on Build GRCh38
rs777915505 Chr.14: 93186636-93186636 on Build GRCh38
rs746775183 Chr.14: 93186645-93186645 on Build GRCh38
rs746614329 Chr.14: 93186530-93186530 on Build GRCh38
rs762504169 Chr.14: 93186695-93186695 on Build GRCh38

View Full Product Details