Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 25175829-25176088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR158
Entrez Gene ID
57512
Gene Name
G protein-coupled receptor 158
Gene Aliases
-
UniGene
Hs.499108
Gene Chromosome Location
Chr.10: 25174722-25602229 on Build GRCh38
Transcripts
NM_020752.2 NR_027333.1

Gene Symbol
GPR158-AS1
Entrez Gene ID
100128811
Gene Name
GPR158 antisense RNA 1
Gene Aliases
-
UniGene
Hs.700407
Gene Chromosome Location
Chr.10: 25158072-25176276 on Build GRCh38
Transcripts
NM_020752.2 NR_027333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5836707 c.520G>A p.A174T Chr.10: 25175940-25175940 on Build GRCh38
4137936 c.573C>A p.F191L Chr.10: 25175993-25175993 on Build GRCh38
4500788 c.573C>T p.F191F Chr.10: 25175993-25175993 on Build GRCh38
5881887 c.539C>T p.T180I Chr.10: 25175959-25175959 on Build GRCh38
7113532 c.587G>T p.R196L Chr.10: 25176007-25176007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551226367 Chr.10: 25175920-25175920 on Build GRCh38
rs754236881 Chr.10: 25175960-25175960 on Build GRCh38
rs774329576 Chr.10: 25176045-25176045 on Build GRCh38
rs767579106 Chr.10: 25176056-25176056 on Build GRCh38
rs758651300 Chr.10: 25176039-25176041 on Build GRCh38
rs773154065 Chr.10: 25175887-25175887 on Build GRCh38
rs754509253 Chr.10: 25175974-25175974 on Build GRCh38
rs756345759 Chr.10: 25175940-25175940 on Build GRCh38
rs535710420 Chr.10: 25175986-25175986 on Build GRCh38
rs746421267 Chr.10: 25175972-25175972 on Build GRCh38
rs930402609 Chr.10: 25175860-25175860 on Build GRCh38
rs865987286 Chr.10: 25176043-25176043 on Build GRCh38
rs773101327 Chr.10: 25176004-25176004 on Build GRCh38
rs1020289295 Chr.10: 25176057-25176057 on Build GRCh38
rs968674168 Chr.10: 25176066-25176066 on Build GRCh38
rs889522425 Chr.10: 25175998-25175998 on Build GRCh38
rs752852266 Chr.10: 25175938-25175938 on Build GRCh38
rs747743861 Chr.10: 25175995-25175995 on Build GRCh38
rs762934432 Chr.10: 25175904-25175904 on Build GRCh38
rs759455582 Chr.10: 25176047-25176047 on Build GRCh38
rs142065649 Chr.10: 25175966-25175966 on Build GRCh38
rs377071000 Chr.10: 25176024-25176024 on Build GRCh38
rs769592452 Chr.10: 25176000-25176000 on Build GRCh38
rs770677082 Chr.10: 25176041-25176041 on Build GRCh38
rs1009896505 Chr.10: 25176036-25176036 on Build GRCh38
rs930985359 Chr.10: 25175951-25175951 on Build GRCh38
rs1040827541 Chr.10: 25176005-25176005 on Build GRCh38
rs779453123 Chr.10: 25175969-25175969 on Build GRCh38
rs372812381 Chr.10: 25175942-25175942 on Build GRCh38
rs747874571 Chr.10: 25175857-25175857 on Build GRCh38
rs61855461 Chr.10: 25175854-25175854 on Build GRCh38
rs749075567 Chr.10: 25176011-25176011 on Build GRCh38
rs940891051 Chr.10: 25175909-25175909 on Build GRCh38
rs905050696 Chr.10: 25175944-25175944 on Build GRCh38
rs918285919 Chr.10: 25175924-25175924 on Build GRCh38
rs199832541 Chr.10: 25175919-25175919 on Build GRCh38
rs370596082 Chr.10: 25175922-25175922 on Build GRCh38
rs769645516 Chr.10: 25175867-25175867 on Build GRCh38

View Full Product Details