Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 170795087-170795360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRP
Entrez Gene ID
2568
Gene Name
gamma-aminobutyric acid type A receptor pi subunit
Gene Aliases
-
UniGene
Hs.26225
Gene Chromosome Location
Chr.5: 170783719-170814047 on Build GRCh38
Transcripts
NM_001291985.1 NM_014211.2 XM_005265872.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1435990 c.244T>C p.Y82H Chr.5: 170795211-170795211 on Build GRCh38
5000974 c.254C>A p.T85N Chr.5: 170795221-170795221 on Build GRCh38
6205745 c.297T>G p.F99L Chr.5: 170795264-170795264 on Build GRCh38
4961440 c.289C>T p.L97L Chr.5: 170795256-170795256 on Build GRCh38
6451719 c.287G>A p.R96Q Chr.5: 170795254-170795254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78994142 Chr.5: 170795191-170795191 on Build GRCh38
rs746589847 Chr.5: 170795328-170795328 on Build GRCh38
rs78320183 Chr.5: 170795190-170795190 on Build GRCh38
rs757616650 Chr.5: 170795251-170795251 on Build GRCh38
rs947407144 Chr.5: 170795173-170795173 on Build GRCh38
rs924601630 Chr.5: 170795226-170795226 on Build GRCh38
rs986241511 Chr.5: 170795157-170795157 on Build GRCh38
rs372934564 Chr.5: 170795312-170795312 on Build GRCh38
rs780404391 Chr.5: 170795324-170795324 on Build GRCh38
rs532076065 Chr.5: 170795218-170795218 on Build GRCh38
rs768154765 Chr.5: 170795200-170795200 on Build GRCh38
rs745452457 Chr.5: 170795179-170795179 on Build GRCh38
rs776531075 Chr.5: 170795201-170795201 on Build GRCh38
rs374054144 Chr.5: 170795271-170795271 on Build GRCh38
rs369574799 Chr.5: 170795300-170795300 on Build GRCh38
rs367942987 Chr.5: 170795272-170795272 on Build GRCh38
rs748020433 Chr.5: 170795276-170795276 on Build GRCh38
rs746391946 Chr.5: 170795253-170795253 on Build GRCh38
rs746470583 Chr.5: 170795262-170795262 on Build GRCh38
rs757722314 Chr.5: 170795334-170795334 on Build GRCh38
rs750747268 Chr.5: 170795320-170795320 on Build GRCh38
rs375800749 Chr.5: 170795332-170795332 on Build GRCh38
rs758944930 Chr.5: 170795322-170795322 on Build GRCh38
rs765321329 Chr.5: 170795223-170795223 on Build GRCh38
rs758887380 Chr.5: 170795171-170795171 on Build GRCh38
rs762916068 Chr.5: 170795233-170795233 on Build GRCh38
rs767719560 Chr.5: 170795236-170795236 on Build GRCh38
rs1013360597 Chr.5: 170795296-170795296 on Build GRCh38
rs866217879 Chr.5: 170795314-170795314 on Build GRCh38
rs903769000 Chr.5: 170795171-170795171 on Build GRCh38
rs910629686 Chr.5: 170795162-170795162 on Build GRCh38
rs758857209 Chr.5: 170795254-170795254 on Build GRCh38
rs536367107 Chr.5: 170795318-170795318 on Build GRCh38
rs754276299 Chr.5: 170795250-170795250 on Build GRCh38
rs368678838 Chr.5: 170795299-170795299 on Build GRCh38
rs779645693 Chr.5: 170795260-170795260 on Build GRCh38
rs199700931 Chr.5: 170795238-170795238 on Build GRCh38
rs763754497 Chr.5: 170795123-170795123 on Build GRCh38
rs771579664 Chr.5: 170795186-170795186 on Build GRCh38
rs563152718 Chr.5: 170795112-170795112 on Build GRCh38
rs773277344 Chr.5: 170795232-170795232 on Build GRCh38
rs77629881 Chr.5: 170795204-170795204 on Build GRCh38
rs771465288 Chr.5: 170795174-170795174 on Build GRCh38
rs942611920 Chr.5: 170795155-170795155 on Build GRCh38
rs181143512 Chr.5: 170795149-170795149 on Build GRCh38
rs749572058 Chr.5: 170795169-170795169 on Build GRCh38
rs779382665 Chr.5: 170795178-170795178 on Build GRCh38
rs764188021 Chr.5: 170795243-170795243 on Build GRCh38
rs754015062 Chr.5: 170795304-170795304 on Build GRCh38
rs779184758 Chr.5: 170795252-170795252 on Build GRCh38
rs773082806 Chr.5: 170795284-170795284 on Build GRCh38
rs756793352 Chr.5: 170795301-170795301 on Build GRCh38
rs41315922 Chr.5: 170795288-170795288 on Build GRCh38
rs186086072 Chr.5: 170795268-170795268 on Build GRCh38
rs774809795 Chr.5: 170795188-170795188 on Build GRCh38
rs145233692 Chr.5: 170795295-170795295 on Build GRCh38
rs912434739 Chr.5: 170795131-170795131 on Build GRCh38
rs775676389 Chr.5: 170795297-170795297 on Build GRCh38
rs149165781 Chr.5: 170795309-170795309 on Build GRCh38
rs149935911 Chr.5: 170795239-170795239 on Build GRCh38

View Full Product Details