Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159262643-159262874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCJ
Entrez Gene ID
654502
Gene Name
IQ motif containing J
Gene Aliases
-
UniGene
Hs.570685
Gene Chromosome Location
Chr.3: 159069252-159266307 on Build GRCh38
Transcripts
NM_001042705.2 NM_001042706.2 NM_001197100.1 NM_001197113.1 NM_001197114.1

Gene Symbol
IQCJ-SCHIP1
Entrez Gene ID
100505385
Gene Name
IQCJ-SCHIP1 readthrough
Gene Aliases
-
UniGene
Hs.134665
Gene Chromosome Location
Chr.3: 159069252-159897366 on Build GRCh38
Transcripts
NM_001042705.2 NM_001042706.2 NM_001197100.1 NM_001197113.1 NM_001197114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3005908 c.277G>A p.D93N Chr.3: 159262669-159262669 on Build GRCh38
368161 c.291C>A p.P97P Chr.3: 159262683-159262683 on Build GRCh38
5708530 c.259A>T p.S87C Chr.3: 159262651-159262651 on Build GRCh38
1040541 c.256G>A p.V86I Chr.3: 159262648-159262648 on Build GRCh38
5708529 c.259A>T p.S87C Chr.3: 159262651-159262651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778714834 Chr.3: 159262693-159262693 on Build GRCh38
rs200957112 Chr.3: 159262669-159262669 on Build GRCh38
rs372523426 Chr.3: 159262739-159262739 on Build GRCh38
rs746491447 Chr.3: 159262719-159262719 on Build GRCh38
rs960627338 Chr.3: 159262801-159262801 on Build GRCh38
rs376824973 Chr.3: 159262746-159262746 on Build GRCh38
rs576185290 Chr.3: 159262799-159262799 on Build GRCh38
rs781259338 Chr.3: 159262677-159262677 on Build GRCh38
rs766893360 Chr.3: 159262668-159262668 on Build GRCh38
rs750756440 Chr.3: 159262665-159262665 on Build GRCh38
rs758785677 Chr.3: 159262708-159262708 on Build GRCh38
rs758844693 Chr.3: 159262667-159262667 on Build GRCh38
rs745684810 Chr.3: 159262704-159262704 on Build GRCh38
rs773576881 Chr.3: 159262773-159262773 on Build GRCh38
rs772098597 Chr.3: 159262705-159262705 on Build GRCh38
rs766850514 Chr.3: 159262776-159262776 on Build GRCh38
rs554739583 Chr.3: 159262707-159262707 on Build GRCh38
rs994510916 Chr.3: 159262690-159262690 on Build GRCh38
rs976476045 Chr.3: 159262697-159262697 on Build GRCh38
rs914755529 Chr.3: 159262805-159262805 on Build GRCh38
rs748470596 Chr.3: 159262682-159262682 on Build GRCh38
rs950430023 Chr.3: 159262737-159262737 on Build GRCh38
rs947512288 Chr.3: 159262826-159262826 on Build GRCh38
rs775875030 Chr.3: 159262778-159262790 on Build GRCh38
rs965483400 Chr.3: 159262684-159262684 on Build GRCh38
rs776140887 Chr.3: 159262745-159262745 on Build GRCh38
rs752118638 Chr.3: 159262780-159262780 on Build GRCh38
rs763338475 Chr.3: 159262774-159262774 on Build GRCh38

View Full Product Details