Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50725043-50725268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC11A
Entrez Gene ID
6320
Gene Name
C-type lectin domain family 11 member A
Gene Aliases
CLECSF3, LSLCL, P47, SCGF
UniGene
Hs.512680
Gene Chromosome Location
Chr.19: 50723348-50725724 on Build GRCh38
Transcripts
NM_002975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316391 c.693C>T p.L231L Chr.19: 50725188-50725188 on Build GRCh38
4795741 c.747C>T p.G249G Chr.19: 50725242-50725242 on Build GRCh38
3389251 c.596C>T p.A199V Chr.19: 50725091-50725091 on Build GRCh38
6046634 c.716G>T p.W239L Chr.19: 50725211-50725211 on Build GRCh38
401879 c.609G>T p.R203R Chr.19: 50725104-50725104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758112465 Chr.19: 50725180-50725180 on Build GRCh38
rs777285246 Chr.19: 50725220-50725220 on Build GRCh38
rs573408703 Chr.19: 50725165-50725165 on Build GRCh38
rs750140748 Chr.19: 50725137-50725137 on Build GRCh38
rs555157998 Chr.19: 50725162-50725162 on Build GRCh38
rs1028530590 Chr.19: 50725060-50725060 on Build GRCh38
rs762153368 Chr.19: 50725176-50725176 on Build GRCh38
rs746583975 Chr.19: 50725144-50725144 on Build GRCh38
rs939120083 Chr.19: 50725093-50725093 on Build GRCh38
rs763352924 Chr.19: 50725088-50725088 on Build GRCh38
rs776262377 Chr.19: 50725083-50725083 on Build GRCh38
rs771250677 Chr.19: 50725089-50725089 on Build GRCh38
rs760857819 Chr.19: 50725171-50725171 on Build GRCh38
rs764239744 Chr.19: 50725174-50725174 on Build GRCh38
rs757159422 Chr.19: 50725132-50725132 on Build GRCh38
rs776249415 Chr.19: 50725081-50725081 on Build GRCh38
rs984419650 Chr.19: 50725134-50725134 on Build GRCh38
rs951722281 Chr.19: 50725078-50725078 on Build GRCh38
rs758372446 Chr.19: 50725138-50725138 on Build GRCh38
rs755766827 Chr.19: 50725213-50725213 on Build GRCh38
rs775848847 Chr.19: 50725169-50725169 on Build GRCh38
rs767852137 Chr.19: 50725115-50725115 on Build GRCh38
rs752825222 Chr.19: 50725123-50725123 on Build GRCh38
rs377437691 Chr.19: 50725133-50725133 on Build GRCh38
rs754091311 Chr.19: 50725126-50725126 on Build GRCh38
rs534274262 Chr.19: 50725166-50725166 on Build GRCh38
rs569727695 Chr.19: 50725109-50725109 on Build GRCh38
rs748078016 Chr.19: 50725087-50725087 on Build GRCh38
rs751417398 Chr.19: 50725192-50725192 on Build GRCh38
rs897021266 Chr.19: 50725097-50725098 on Build GRCh38
rs958263363 Chr.19: 50725072-50725072 on Build GRCh38
rs771592326 Chr.19: 50725113-50725113 on Build GRCh38
rs939980763 Chr.19: 50725151-50725151 on Build GRCh38
rs772387186 Chr.19: 50725080-50725080 on Build GRCh38
rs765501319 Chr.19: 50725177-50725177 on Build GRCh38
rs766385763 Chr.19: 50725188-50725188 on Build GRCh38
rs951691221 Chr.19: 50725172-50725172 on Build GRCh38
rs780859030 Chr.19: 50725160-50725160 on Build GRCh38
rs779791365 Chr.19: 50725140-50725140 on Build GRCh38
rs747826397 Chr.19: 50725161-50725161 on Build GRCh38
rs776868541 Chr.19: 50725241-50725241 on Build GRCh38
rs765429884 Chr.19: 50725089-50725090 on Build GRCh38
rs761771291 Chr.19: 50725124-50725124 on Build GRCh38
rs747600004 Chr.19: 50725207-50725207 on Build GRCh38
rs367979493 Chr.19: 50725243-50725243 on Build GRCh38
rs769240488 Chr.19: 50725239-50725239 on Build GRCh38
rs903849798 Chr.19: 50725129-50725129 on Build GRCh38
rs137983631 Chr.19: 50725224-50725224 on Build GRCh38
rs769843229 Chr.19: 50725153-50725153 on Build GRCh38
rs774657810 Chr.19: 50725104-50725104 on Build GRCh38
rs562569155 Chr.19: 50725114-50725115 on Build GRCh38
rs907658544 Chr.19: 50725092-50725092 on Build GRCh38
rs754867017 Chr.19: 50725197-50725197 on Build GRCh38
rs201049088 Chr.19: 50725070-50725070 on Build GRCh38
rs747550305 Chr.19: 50725232-50725232 on Build GRCh38
rs537832690 Chr.19: 50725240-50725240 on Build GRCh38
rs750240450 Chr.19: 50725179-50725179 on Build GRCh38
rs977124168 Chr.19: 50725116-50725116 on Build GRCh38
rs758469284 Chr.19: 50725166-50725189 on Build GRCh38
rs370253740 Chr.19: 50725158-50725158 on Build GRCh38
rs764470824 Chr.19: 50725125-50725125 on Build GRCh38
rs991273108 Chr.19: 50725131-50725131 on Build GRCh38
rs759751080 Chr.19: 50725168-50725168 on Build GRCh38
rs201753001 Chr.19: 50725178-50725178 on Build GRCh38
rs780701862 Chr.19: 50725204-50725204 on Build GRCh38

View Full Product Details