Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23559082-23559307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ID3
Entrez Gene ID
3399
Gene Name
inhibitor of DNA binding 3, HLH protein
Gene Aliases
HEIR-1, bHLHb25
UniGene
Hs.76884
Gene Chromosome Location
Chr.1: 23557930-23559794 on Build GRCh38
Transcripts
NM_002167.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
382960 c.235G>T p.D79Y Chr.1: 23559192-23559192 on Build GRCh38
6221985 c.300+10G>T p.? Chr.1: 23559117-23559117 on Build GRCh38
2049209 c.208C>G p.L70V Chr.1: 23559219-23559219 on Build GRCh38
6211071 c.167C>A p.P56H Chr.1: 23559260-23559260 on Build GRCh38
5652077 c.237C>T p.D79D Chr.1: 23559190-23559190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139428797 Chr.1: 23559172-23559172 on Build GRCh38
rs146830460 Chr.1: 23559247-23559247 on Build GRCh38
rs776260530 Chr.1: 23559117-23559117 on Build GRCh38
rs201313484 Chr.1: 23559160-23559160 on Build GRCh38
rs753164192 Chr.1: 23559222-23559222 on Build GRCh38
rs781698505 Chr.1: 23559163-23559163 on Build GRCh38
rs764670040 Chr.1: 23559195-23559195 on Build GRCh38
rs1055074154 Chr.1: 23559237-23559237 on Build GRCh38
rs148996867 Chr.1: 23559183-23559183 on Build GRCh38
rs570232356 Chr.1: 23559277-23559277 on Build GRCh38
rs768684566 Chr.1: 23559190-23559190 on Build GRCh38
rs772857805 Chr.1: 23559200-23559200 on Build GRCh38
rs61749352 Chr.1: 23559171-23559171 on Build GRCh38
rs528089188 Chr.1: 23559135-23559135 on Build GRCh38
rs761339096 Chr.1: 23559193-23559193 on Build GRCh38
rs766082863 Chr.1: 23559205-23559205 on Build GRCh38
rs759930189 Chr.1: 23559212-23559212 on Build GRCh38
rs775420234 Chr.1: 23559179-23559179 on Build GRCh38
rs764184904 Chr.1: 23559111-23559111 on Build GRCh38
rs761271826 Chr.1: 23559118-23559118 on Build GRCh38
rs2067053 Chr.1: 23559108-23559108 on Build GRCh38
rs762512538 Chr.1: 23559204-23559204 on Build GRCh38
rs11542319 Chr.1: 23559211-23559211 on Build GRCh38
rs150987910 Chr.1: 23559259-23559259 on Build GRCh38
rs767969316 Chr.1: 23559217-23559217 on Build GRCh38
rs748044061 Chr.1: 23559267-23559267 on Build GRCh38
rs748225151 Chr.1: 23559164-23559164 on Build GRCh38
rs370478639 Chr.1: 23559169-23559169 on Build GRCh38
rs766303297 Chr.1: 23559151-23559151 on Build GRCh38
rs758735312 Chr.1: 23559145-23559145 on Build GRCh38
rs776741657 Chr.1: 23559192-23559192 on Build GRCh38
rs146163818 Chr.1: 23559282-23559282 on Build GRCh38
rs934808402 Chr.1: 23559229-23559229 on Build GRCh38
rs148870313 Chr.1: 23559250-23559250 on Build GRCh38
rs11542318 Chr.1: 23559136-23559136 on Build GRCh38
rs767879343 Chr.1: 23559286-23559286 on Build GRCh38
rs374890938 Chr.1: 23559281-23559281 on Build GRCh38
rs202157091 Chr.1: 23559251-23559251 on Build GRCh38
rs972563455 Chr.1: 23559182-23559182 on Build GRCh38
rs772588913 Chr.1: 23559274-23559274 on Build GRCh38
rs140027264 Chr.1: 23559226-23559226 on Build GRCh38
rs910526315 Chr.1: 23559187-23559187 on Build GRCh38
rs764905305 Chr.1: 23559131-23559131 on Build GRCh38
rs969265582 Chr.1: 23559153-23559153 on Build GRCh38
rs201168120 Chr.1: 23559150-23559150 on Build GRCh38
rs1023747415 Chr.1: 23559149-23559149 on Build GRCh38
rs367596122 Chr.1: 23559287-23559287 on Build GRCh38
rs377603935 Chr.1: 23559109-23559109 on Build GRCh38
rs1010571884 Chr.1: 23559144-23559144 on Build GRCh38
rs770537277 Chr.1: 23559279-23559279 on Build GRCh38
rs377385656 Chr.1: 23559245-23559245 on Build GRCh38
rs1034901687 Chr.1: 23559156-23559156 on Build GRCh38
rs147380623 Chr.1: 23559155-23559155 on Build GRCh38
rs777718207 Chr.1: 23559228-23559228 on Build GRCh38
rs11542320 Chr.1: 23559142-23559142 on Build GRCh38
rs754016671 Chr.1: 23559238-23559238 on Build GRCh38
rs769841287 Chr.1: 23559272-23559272 on Build GRCh38
rs560971527 Chr.1: 23559110-23559110 on Build GRCh38
rs768596654 Chr.1: 23559258-23559258 on Build GRCh38
rs997650130 Chr.1: 23559106-23559106 on Build GRCh38

View Full Product Details