Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135345873-135346146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFY
Entrez Gene ID
9555
Gene Name
H2A histone family member Y
Gene Aliases
H2A.y, H2A/y, H2AF12M, MACROH2A1.1, mH2A1, macroH2A1.2
UniGene
Hs.420272
Gene Chromosome Location
Chr.5: 135334381-135399887 on Build GRCh38
Transcripts
NM_001040158.1 NM_004893.2 NM_138609.2 NM_138610.2 XM_005272132.1 XM_011543729.2 XM_011543730.2 XM_011543731.1 XM_011543732.1 XM_011543733.2 XM_017010067.1 XM_017010069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060984 c.744G>T p.K248N Chr.5: 135345993-135345993 on Build GRCh38
5432814 c.763T>G p.L255V Chr.5: 135345983-135345983 on Build GRCh38
3850225 c.778G>A p.A260T Chr.5: 135345968-135345968 on Build GRCh38
3850226 c.769G>A p.A257T Chr.5: 135345968-135345968 on Build GRCh38
5432815 c.754T>G p.L252V Chr.5: 135345983-135345983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538418903 Chr.5: 135345901-135345901 on Build GRCh38
rs748611698 Chr.5: 135345936-135345936 on Build GRCh38
rs748616415 Chr.5: 135346071-135346071 on Build GRCh38
rs773423763 Chr.5: 135345938-135345938 on Build GRCh38
rs553875930 Chr.5: 135345924-135345924 on Build GRCh38
rs924380331 Chr.5: 135345920-135345920 on Build GRCh38
rs944014626 Chr.5: 135346068-135346068 on Build GRCh38
rs751159474 Chr.5: 135346021-135346021 on Build GRCh38
rs775443375 Chr.5: 135346104-135346104 on Build GRCh38
rs1038335125 Chr.5: 135346017-135346017 on Build GRCh38
rs147522551 Chr.5: 135346102-135346102 on Build GRCh38
rs780802411 Chr.5: 135346038-135346038 on Build GRCh38
rs762867922 Chr.5: 135345994-135345994 on Build GRCh38
rs1007766259 Chr.5: 135345986-135345986 on Build GRCh38
rs377698309 Chr.5: 135346073-135346073 on Build GRCh38
rs752434135 Chr.5: 135345990-135345990 on Build GRCh38
rs376197418 Chr.5: 135346013-135346013 on Build GRCh38
rs572189889 Chr.5: 135345937-135345937 on Build GRCh38
rs371223386 Chr.5: 135346088-135346088 on Build GRCh38
rs771187482 Chr.5: 135346099-135346099 on Build GRCh38
rs1051913001 Chr.5: 135345932-135345932 on Build GRCh38
rs143164737 Chr.5: 135346101-135346101 on Build GRCh38
rs779125489 Chr.5: 135345925-135345925 on Build GRCh38
rs760811398 Chr.5: 135345949-135345949 on Build GRCh38
rs776711803 Chr.5: 135345958-135345958 on Build GRCh38
rs542695907 Chr.5: 135345957-135345957 on Build GRCh38
rs374137994 Chr.5: 135346094-135346094 on Build GRCh38
rs1051680311 Chr.5: 135346000-135346000 on Build GRCh38
rs765195054 Chr.5: 135345989-135345989 on Build GRCh38
rs757002605 Chr.5: 135346023-135346023 on Build GRCh38
rs759443476 Chr.5: 135345971-135345971 on Build GRCh38
rs750731138 Chr.5: 135345923-135345923 on Build GRCh38
rs750002834 Chr.5: 135346045-135346045 on Build GRCh38
rs978508339 Chr.5: 135345948-135345948 on Build GRCh38
rs755511853 Chr.5: 135346050-135346050 on Build GRCh38
rs749775785 Chr.5: 135345919-135345919 on Build GRCh38
rs554945747 Chr.5: 135346081-135346081 on Build GRCh38

View Full Product Details