Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222303801-222304062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC140
Entrez Gene ID
151278
Gene Name
coiled-coil domain containing 140
Gene Aliases
-
UniGene
Hs.350729
Gene Chromosome Location
Chr.2: 222298147-222305217 on Build GRCh38
Transcripts
NM_153038.1 XM_017003466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1291451 c.33C>A p.L11L Chr.2: 222303935-222303935 on Build GRCh38
3730598 c.17C>A p.S6* Chr.2: 222303919-222303919 on Build GRCh38
1306481 c.90G>A p.A30A Chr.2: 222303992-222303992 on Build GRCh38
1405798 c.131A>G p.H44R Chr.2: 222304033-222304033 on Build GRCh38
7359773 c.104C>A p.T35N Chr.2: 222304006-222304006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555229091 Chr.2: 222303840-222303840 on Build GRCh38
rs960481002 Chr.2: 222303841-222303841 on Build GRCh38
rs201194085 Chr.2: 222304039-222304039 on Build GRCh38
rs936851862 Chr.2: 222303834-222303834 on Build GRCh38
rs540295380 Chr.2: 222303924-222303924 on Build GRCh38
rs770620395 Chr.2: 222303941-222303941 on Build GRCh38
rs144424153 Chr.2: 222303887-222303887 on Build GRCh38
rs780220477 Chr.2: 222303881-222303881 on Build GRCh38
rs756237142 Chr.2: 222303996-222303996 on Build GRCh38
rs752719755 Chr.2: 222303963-222303963 on Build GRCh38
rs767760565 Chr.2: 222303973-222303973 on Build GRCh38
rs759395793 Chr.2: 222303952-222303952 on Build GRCh38
rs1022650237 Chr.2: 222304003-222304003 on Build GRCh38
rs1050446648 Chr.2: 222303986-222303986 on Build GRCh38
rs185091025 Chr.2: 222303984-222303984 on Build GRCh38
rs776229705 Chr.2: 222303946-222303946 on Build GRCh38
rs770553672 Chr.2: 222303930-222303932 on Build GRCh38
rs888348460 Chr.2: 222303896-222303896 on Build GRCh38
rs780349625 Chr.2: 222304004-222304004 on Build GRCh38
rs923299225 Chr.2: 222303974-222303974 on Build GRCh38
rs765224879 Chr.2: 222303956-222303956 on Build GRCh38
rs976106483 Chr.2: 222303968-222303968 on Build GRCh38
rs535377834 Chr.2: 222303837-222303837 on Build GRCh38
rs749623288 Chr.2: 222304037-222304037 on Build GRCh38
rs549681845 Chr.2: 222303995-222303995 on Build GRCh38
rs925549838 Chr.2: 222304031-222304031 on Build GRCh38
rs199997238 Chr.2: 222303861-222303861 on Build GRCh38
rs756483844 Chr.2: 222303880-222303880 on Build GRCh38
rs147196537 Chr.2: 222303909-222303909 on Build GRCh38
rs140701501 Chr.2: 222304008-222304008 on Build GRCh38
rs781531888 Chr.2: 222303863-222303863 on Build GRCh38
rs772838123 Chr.2: 222303916-222303916 on Build GRCh38
rs1052587850 Chr.2: 222303838-222303838 on Build GRCh38
rs754115382 Chr.2: 222304005-222304005 on Build GRCh38
rs568741324 Chr.2: 222303845-222303845 on Build GRCh38
rs755378978 Chr.2: 222304006-222304006 on Build GRCh38
rs771567540 Chr.2: 222304017-222304017 on Build GRCh38
rs959792438 Chr.2: 222303954-222303954 on Build GRCh38
rs944095175 Chr.2: 222303846-222303846 on Build GRCh38
rs750656604 Chr.2: 222303869-222303869 on Build GRCh38
rs761959217 Chr.2: 222303969-222303969 on Build GRCh38
rs779429538 Chr.2: 222303898-222303898 on Build GRCh38
rs958274944 Chr.2: 222304038-222304038 on Build GRCh38
rs78734419 Chr.2: 222303856-222303856 on Build GRCh38
rs777877627 Chr.2: 222304010-222304011 on Build GRCh38
rs751515543 Chr.2: 222303906-222303906 on Build GRCh38
rs78030705 Chr.2: 222303976-222303976 on Build GRCh38
rs769106003 Chr.2: 222303894-222303894 on Build GRCh38
rs779972404 Chr.2: 222303892-222303892 on Build GRCh38
rs771620847 Chr.2: 222303914-222303914 on Build GRCh38
rs533040478 Chr.2: 222303975-222303975 on Build GRCh38
rs751639272 Chr.2: 222303858-222303858 on Build GRCh38

View Full Product Details