Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68761482-68761739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT1A
Entrez Gene ID
1374
Gene Name
carnitine palmitoyltransferase 1A
Gene Aliases
CPT1, CPT1-L, L-CPT1
UniGene
Hs.503043
Gene Chromosome Location
Chr.11: 68754620-68844410 on Build GRCh38
Transcripts
NM_001031847.2 NM_001876.3 XM_005273762.2 XM_005273763.1 XM_017017220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2042963 c.1934G>A p.R645H Chr.11: 68761629-68761629 on Build GRCh38
2042961 c.1963C>T p.R655C Chr.11: 68761600-68761600 on Build GRCh38
6134444 c.1949G>T p.G650V Chr.11: 68761614-68761614 on Build GRCh38
5611973 c.1896G>A p.L632L Chr.11: 68761667-68761667 on Build GRCh38
6294328 c.1946C>A p.T649N Chr.11: 68761617-68761617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80356799 Chr.11: 68761533-68761536 on Build GRCh38
rs768465007 Chr.11: 68761528-68761528 on Build GRCh38
rs760769734 Chr.11: 68761615-68761615 on Build GRCh38
rs1044748249 Chr.11: 68761675-68761675 on Build GRCh38
rs556392764 Chr.11: 68761604-68761604 on Build GRCh38
rs775438656 Chr.11: 68761656-68761656 on Build GRCh38
rs537749330 Chr.11: 68761549-68761549 on Build GRCh38
rs75908421 Chr.11: 68761543-68761543 on Build GRCh38
rs375814998 Chr.11: 68761636-68761636 on Build GRCh38
rs2228503 Chr.11: 68761559-68761559 on Build GRCh38
rs755128170 Chr.11: 68761621-68761621 on Build GRCh38
rs766536084 Chr.11: 68761616-68761616 on Build GRCh38
rs2228503 Chr.11: 68761559-68761559 on Build GRCh38
rs754141480 Chr.11: 68761618-68761618 on Build GRCh38
rs768746064 Chr.11: 68761686-68761686 on Build GRCh38
rs111407620 Chr.11: 68761655-68761655 on Build GRCh38
rs374136875 Chr.11: 68761580-68761580 on Build GRCh38
rs776573287 Chr.11: 68761716-68761716 on Build GRCh38
rs755323437 Chr.11: 68761550-68761550 on Build GRCh38
rs752632240 Chr.11: 68761513-68761513 on Build GRCh38
rs756306465 Chr.11: 68761532-68761532 on Build GRCh38
rs754188942 Chr.11: 68761548-68761548 on Build GRCh38
rs758916498 Chr.11: 68761624-68761624 on Build GRCh38
rs750518086 Chr.11: 68761529-68761529 on Build GRCh38
rs780257705 Chr.11: 68761546-68761546 on Build GRCh38
rs1057516800 Chr.11: 68761565-68761566 on Build GRCh38
rs375809779 Chr.11: 68761561-68761561 on Build GRCh38
rs1006087779 Chr.11: 68761702-68761702 on Build GRCh38
rs777319252 Chr.11: 68761574-68761574 on Build GRCh38
rs1015556608 Chr.11: 68761713-68761713 on Build GRCh38
rs776306063 Chr.11: 68761579-68761579 on Build GRCh38
rs115731492 Chr.11: 68761622-68761622 on Build GRCh38
rs1042056896 Chr.11: 68761619-68761619 on Build GRCh38
rs552887404 Chr.11: 68761710-68761710 on Build GRCh38
rs368239019 Chr.11: 68761639-68761639 on Build GRCh38
rs746455686 Chr.11: 68761576-68761576 on Build GRCh38
rs994056476 Chr.11: 68761682-68761682 on Build GRCh38
rs762960956 Chr.11: 68761523-68761523 on Build GRCh38
rs367879657 Chr.11: 68761704-68761704 on Build GRCh38
rs372215145 Chr.11: 68761660-68761660 on Build GRCh38
rs569816517 Chr.11: 68761605-68761605 on Build GRCh38
rs748821855 Chr.11: 68761513-68761513 on Build GRCh38
rs763042338 Chr.11: 68761600-68761600 on Build GRCh38
rs41302429 Chr.11: 68761687-68761687 on Build GRCh38
rs370172029 Chr.11: 68761562-68761562 on Build GRCh38
rs371411802 Chr.11: 68761625-68761625 on Build GRCh38
rs369331378 Chr.11: 68761512-68761512 on Build GRCh38
rs376959593 Chr.11: 68761599-68761599 on Build GRCh38
rs150459546 Chr.11: 68761653-68761653 on Build GRCh38
rs769679090 Chr.11: 68761592-68761592 on Build GRCh38
rs367840385 Chr.11: 68761630-68761630 on Build GRCh38
rs80356798 Chr.11: 68761688-68761688 on Build GRCh38
rs770491096 Chr.11: 68761578-68761578 on Build GRCh38
rs761635996 Chr.11: 68761693-68761693 on Build GRCh38

View Full Product Details