Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143726631-143726878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83H
Entrez Gene ID
286077
Gene Name
family with sequence similarity 83 member H
Gene Aliases
AI3
UniGene
Hs.67776
Gene Chromosome Location
Chr.8: 143723933-143739947 on Build GRCh38
Transcripts
NM_198488.3 XM_005250887.3 XM_005250888.3 XM_005250889.3 XM_011516980.2 XM_011516981.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355782 c.2610C>T p.S870S Chr.8: 143726851-143726851 on Build GRCh38
6856503 c.2787delC p.V930fs*44 Chr.8: 143726674-143726674 on Build GRCh38
239810 c.2797C>T p.R933C Chr.8: 143726664-143726664 on Build GRCh38
6840504 c.2765G>A p.R922H Chr.8: 143726696-143726696 on Build GRCh38
5544972 c.2750C>T p.P917L Chr.8: 143726711-143726711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116851527 Chr.8: 143726756-143726756 on Build GRCh38
rs782198800 Chr.8: 143726740-143726740 on Build GRCh38
rs782777192 Chr.8: 143726704-143726704 on Build GRCh38
rs200179667 Chr.8: 143726783-143726783 on Build GRCh38
rs782554162 Chr.8: 143726850-143726850 on Build GRCh38
rs781957914 Chr.8: 143726701-143726701 on Build GRCh38
rs782030376 Chr.8: 143726707-143726707 on Build GRCh38
rs782548156 Chr.8: 143726733-143726733 on Build GRCh38
rs781886472 Chr.8: 143726848-143726848 on Build GRCh38
rs370284848 Chr.8: 143726668-143726668 on Build GRCh38
rs372449536 Chr.8: 143726841-143726841 on Build GRCh38
rs928984188 Chr.8: 143726689-143726689 on Build GRCh38
rs782743196 Chr.8: 143726780-143726780 on Build GRCh38
rs548161428 Chr.8: 143726684-143726684 on Build GRCh38
rs782294025 Chr.8: 143726674-143726674 on Build GRCh38
rs782632537 Chr.8: 143726673-143726673 on Build GRCh38
rs782289879 Chr.8: 143726762-143726762 on Build GRCh38
rs114041611 Chr.8: 143726836-143726836 on Build GRCh38
rs782330086 Chr.8: 143726837-143726837 on Build GRCh38
rs201552119 Chr.8: 143726828-143726828 on Build GRCh38
rs782814594 Chr.8: 143726786-143726786 on Build GRCh38
rs782691731 Chr.8: 143726801-143726801 on Build GRCh38
rs781782862 Chr.8: 143726719-143726719 on Build GRCh38
rs202016228 Chr.8: 143726652-143726652 on Build GRCh38
rs576698235 Chr.8: 143726835-143726835 on Build GRCh38
rs781924646 Chr.8: 143726840-143726840 on Build GRCh38
rs781831914 Chr.8: 143726854-143726854 on Build GRCh38
rs782123049 Chr.8: 143726785-143726785 on Build GRCh38
rs906265524 Chr.8: 143726708-143726708 on Build GRCh38
rs782187271 Chr.8: 143726686-143726686 on Build GRCh38
rs558503828 Chr.8: 143726724-143726724 on Build GRCh38
rs782681351 Chr.8: 143726653-143726653 on Build GRCh38
rs782476188 Chr.8: 143726810-143726810 on Build GRCh38
rs200164567 Chr.8: 143726691-143726691 on Build GRCh38
rs782210087 Chr.8: 143726829-143726829 on Build GRCh38
rs782399921 Chr.8: 143726669-143726669 on Build GRCh38
rs199730760 Chr.8: 143726816-143726816 on Build GRCh38
rs781817049 Chr.8: 143726757-143726757 on Build GRCh38
rs781893815 Chr.8: 143726794-143726794 on Build GRCh38
rs782412107 Chr.8: 143726679-143726679 on Build GRCh38
rs781934743 Chr.8: 143726767-143726767 on Build GRCh38
rs782674498 Chr.8: 143726817-143726817 on Build GRCh38
rs782682699 Chr.8: 143726736-143726736 on Build GRCh38
rs782448672 Chr.8: 143726663-143726663 on Build GRCh38
rs782471542 Chr.8: 143726800-143726800 on Build GRCh38
rs374727482 Chr.8: 143726845-143726845 on Build GRCh38
rs202057687 Chr.8: 143726823-143726823 on Build GRCh38
rs782441160 Chr.8: 143726742-143726742 on Build GRCh38
rs554945877 Chr.8: 143726834-143726834 on Build GRCh38
rs782246101 Chr.8: 143726813-143726813 on Build GRCh38
rs782125910 Chr.8: 143726702-143726702 on Build GRCh38
rs782143259 Chr.8: 143726710-143726710 on Build GRCh38
rs537326894 Chr.8: 143726722-143726722 on Build GRCh38
rs782400915 Chr.8: 143726763-143726763 on Build GRCh38
rs781884876 Chr.8: 143726662-143726662 on Build GRCh38
rs782455374 Chr.8: 143726849-143726849 on Build GRCh38
rs534779867 Chr.8: 143726753-143726753 on Build GRCh38
rs781931295 Chr.8: 143726673-143726674 on Build GRCh38
rs954767327 Chr.8: 143726796-143726796 on Build GRCh38
rs782044744 Chr.8: 143726770-143726770 on Build GRCh38
rs781981150 Chr.8: 143726758-143726758 on Build GRCh38
rs431825180 Chr.8: 143726696-143726696 on Build GRCh38
rs782361452 Chr.8: 143726697-143726697 on Build GRCh38
rs920214848 Chr.8: 143726660-143726660 on Build GRCh38
rs781840200 Chr.8: 143726808-143726808 on Build GRCh38
rs138078852 Chr.8: 143726725-143726725 on Build GRCh38
rs367818194 Chr.8: 143726713-143726713 on Build GRCh38
rs431825181 Chr.8: 143726695-143726695 on Build GRCh38
rs782190692 Chr.8: 143726819-143726819 on Build GRCh38

View Full Product Details