Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73449427-73449694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ1B
Entrez Gene ID
9031
Gene Name
bromodomain adjacent to zinc finger domain 1B
Gene Aliases
WBSCR10, WBSCR9, WSTF
UniGene
Hs.743372
Gene Chromosome Location
Chr.7: 73440398-73522298 on Build GRCh38
Transcripts
NM_032408.3 XM_017012773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5455597 c.3581G>A p.G1194D Chr.7: 73449689-73449689 on Build GRCh38
3640343 c.3720C>T p.S1240S Chr.7: 73449550-73449550 on Build GRCh38
1184476 c.3713G>A p.R1238H Chr.7: 73449557-73449557 on Build GRCh38
5445446 c.3627C>T p.F1209F Chr.7: 73449643-73449643 on Build GRCh38
3882234 c.3621A>C p.K1207N Chr.7: 73449649-73449649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782727805 Chr.7: 73449500-73449500 on Build GRCh38
rs782815047 Chr.7: 73449542-73449542 on Build GRCh38
rs956106745 Chr.7: 73449470-73449470 on Build GRCh38
rs191846102 Chr.7: 73449488-73449488 on Build GRCh38
rs369786974 Chr.7: 73449549-73449549 on Build GRCh38
rs782039360 Chr.7: 73449537-73449537 on Build GRCh38
rs782066365 Chr.7: 73449620-73449620 on Build GRCh38
rs782273444 Chr.7: 73449548-73449548 on Build GRCh38
rs782290096 Chr.7: 73449565-73449565 on Build GRCh38
rs370466076 Chr.7: 73449600-73449600 on Build GRCh38
rs782576967 Chr.7: 73449559-73449559 on Build GRCh38
rs368673994 Chr.7: 73449491-73449491 on Build GRCh38
rs781809533 Chr.7: 73449527-73449527 on Build GRCh38
rs782661604 Chr.7: 73449586-73449586 on Build GRCh38
rs782624497 Chr.7: 73449551-73449551 on Build GRCh38
rs909763054 Chr.7: 73449609-73449609 on Build GRCh38
rs782404725 Chr.7: 73449571-73449571 on Build GRCh38
rs375694776 Chr.7: 73449558-73449558 on Build GRCh38
rs782312401 Chr.7: 73449643-73449643 on Build GRCh38
rs781831625 Chr.7: 73449554-73449554 on Build GRCh38
rs782466767 Chr.7: 73449529-73449529 on Build GRCh38
rs782354690 Chr.7: 73449606-73449606 on Build GRCh38
rs372634828 Chr.7: 73449557-73449557 on Build GRCh38
rs782098674 Chr.7: 73449494-73449494 on Build GRCh38
rs781952325 Chr.7: 73449607-73449607 on Build GRCh38

View Full Product Details