Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48074933-48075176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX1
Entrez Gene ID
10951
Gene Name
chromobox 1
Gene Aliases
CBX, HP1-BETA, HP1Hs-beta, HP1Hsbeta, M31, MOD1, p25beta
UniGene
Hs.77254
Gene Chromosome Location
Chr.17: 48070052-48101521 on Build GRCh38
Transcripts
NM_001127228.1 NM_006807.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2697272 c.354G>A p.E118E Chr.17: 48075065-48075065 on Build GRCh38
980585 c.394_399delCTCATG p.L132_M133delLM Chr.17: 48075020-48075025 on Build GRCh38
436777 c.358G>A p.E120K Chr.17: 48075061-48075061 on Build GRCh38
6621006 c.331C>T p.R111* Chr.17: 48075088-48075088 on Build GRCh38
3795724 c.371G>C p.G124A Chr.17: 48075048-48075048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370438781 Chr.17: 48074975-48074975 on Build GRCh38
rs774718299 Chr.17: 48074966-48074966 on Build GRCh38
rs762176192 Chr.17: 48075143-48075143 on Build GRCh38
rs2240121 Chr.17: 48074960-48074960 on Build GRCh38
rs184622224 Chr.17: 48075137-48075137 on Build GRCh38
rs551242576 Chr.17: 48075062-48075062 on Build GRCh38
rs928210845 Chr.17: 48075017-48075017 on Build GRCh38
rs200328340 Chr.17: 48074969-48074969 on Build GRCh38
rs547942391 Chr.17: 48074977-48074977 on Build GRCh38
rs776515108 Chr.17: 48074968-48074969 on Build GRCh38
rs559113773 Chr.17: 48074970-48074970 on Build GRCh38
rs753983784 Chr.17: 48074969-48074971 on Build GRCh38
rs780623608 Chr.17: 48075113-48075113 on Build GRCh38
rs367599145 Chr.17: 48074999-48074999 on Build GRCh38
rs1011065104 Chr.17: 48075147-48075147 on Build GRCh38
rs762120620 Chr.17: 48074968-48074968 on Build GRCh38
rs749690535 Chr.17: 48075118-48075118 on Build GRCh38
rs777301307 Chr.17: 48075023-48075023 on Build GRCh38
rs774845193 Chr.17: 48075141-48075141 on Build GRCh38
rs893974270 Chr.17: 48075123-48075123 on Build GRCh38
rs1050122249 Chr.17: 48075154-48075154 on Build GRCh38
rs746549679 Chr.17: 48075035-48075035 on Build GRCh38
rs1052516536 Chr.17: 48075121-48075121 on Build GRCh38
rs191768236 Chr.17: 48074993-48074993 on Build GRCh38
rs140625114 Chr.17: 48074998-48074998 on Build GRCh38
rs753674268 Chr.17: 48074978-48074978 on Build GRCh38
rs768843643 Chr.17: 48074965-48074965 on Build GRCh38
rs2875747 Chr.17: 48074971-48074971 on Build GRCh38
rs775816238 Chr.17: 48074958-48074958 on Build GRCh38
rs761694837 Chr.17: 48074969-48074969 on Build GRCh38
rs984194830 Chr.17: 48074988-48074988 on Build GRCh38
rs768725433 Chr.17: 48074967-48074968 on Build GRCh38
rs769530112 Chr.17: 48074975-48074977 on Build GRCh38
rs968341788 Chr.17: 48075140-48075140 on Build GRCh38
rs772908393 Chr.17: 48075120-48075121 on Build GRCh38

View Full Product Details