Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28483139-28483380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO16
Entrez Gene ID
157574
Gene Name
F-box protein 16
Gene Aliases
FBX16
UniGene
Hs.741522
Gene Chromosome Location
Chr.8: 28428408-28490318 on Build GRCh38
Transcripts
NM_001258211.1 NM_172366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874522 c.77A>C p.N26T Chr.8: 28483370-28483370 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753261495 Chr.8: 28483297-28483297 on Build GRCh38
rs146898385 Chr.8: 28483259-28483260 on Build GRCh38
rs533293936 Chr.8: 28483239-28483239 on Build GRCh38
rs779880602 Chr.8: 28483336-28483336 on Build GRCh38
rs985523455 Chr.8: 28483248-28483248 on Build GRCh38
rs140479126 Chr.8: 28483281-28483281 on Build GRCh38
rs912268900 Chr.8: 28483260-28483260 on Build GRCh38
rs368926713 Chr.8: 28483322-28483322 on Build GRCh38
rs371356262 Chr.8: 28483337-28483337 on Build GRCh38
rs761794269 Chr.8: 28483349-28483349 on Build GRCh38
rs768441189 Chr.8: 28483340-28483340 on Build GRCh38
rs373752408 Chr.8: 28483348-28483348 on Build GRCh38
rs756207315 Chr.8: 28483303-28483303 on Build GRCh38
rs375006428 Chr.8: 28483305-28483305 on Build GRCh38
rs992863191 Chr.8: 28483275-28483275 on Build GRCh38
rs551237917 Chr.8: 28483213-28483213 on Build GRCh38
rs973962238 Chr.8: 28483183-28483183 on Build GRCh38
rs904123807 Chr.8: 28483295-28483295 on Build GRCh38
rs927392343 Chr.8: 28483244-28483244 on Build GRCh38
rs75164855 Chr.8: 28483261-28483261 on Build GRCh38
rs377600824 Chr.8: 28483350-28483350 on Build GRCh38
rs1052903046 Chr.8: 28483268-28483268 on Build GRCh38
rs543247554 Chr.8: 28483247-28483247 on Build GRCh38
rs1034076634 Chr.8: 28483339-28483339 on Build GRCh38
rs749524525 Chr.8: 28483314-28483314 on Build GRCh38
rs1043919727 Chr.8: 28483291-28483291 on Build GRCh38
rs777713130 Chr.8: 28483200-28483200 on Build GRCh38

View Full Product Details