Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101753317-101753580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTAB
Entrez Gene ID
79158
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Gene Aliases
GNPTA, ICD
UniGene
Hs.46850
Gene Chromosome Location
Chr.12: 101745497-101830867 on Build GRCh38
Transcripts
NM_024312.4 XM_011538731.2 XM_017019961.1 XM_017019962.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672452 c.3440T>G p.F1147C Chr.12: 101753534-101753534 on Build GRCh38
4490114 c.3580C>T p.H1194Y Chr.12: 101753394-101753394 on Build GRCh38
934034 c.3589G>A p.E1197K Chr.12: 101753385-101753385 on Build GRCh38
7179111 c.3559G>A p.E1187K Chr.12: 101753415-101753415 on Build GRCh38
1706130 c.3576C>A p.F1192L Chr.12: 101753398-101753398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755389623 Chr.12: 101753464-101753464 on Build GRCh38
rs34002892 Chr.12: 101753470-101753471 on Build GRCh38
rs773382436 Chr.12: 101753541-101753542 on Build GRCh38
rs137852897 Chr.12: 101753409-101753409 on Build GRCh38
rs137853825 Chr.12: 101753376-101753376 on Build GRCh38
rs770515265 Chr.12: 101753442-101753442 on Build GRCh38
rs558234007 Chr.12: 101753549-101753549 on Build GRCh38
rs766234122 Chr.12: 101753348-101753348 on Build GRCh38
rs777256692 Chr.12: 101753434-101753434 on Build GRCh38
rs775626564 Chr.12: 101753450-101753450 on Build GRCh38
rs587777913 Chr.12: 101753406-101753407 on Build GRCh38
rs750709036 Chr.12: 101753383-101753383 on Build GRCh38
rs369510134 Chr.12: 101753446-101753446 on Build GRCh38
rs765449372 Chr.12: 101753456-101753456 on Build GRCh38
rs141007019 Chr.12: 101753408-101753408 on Build GRCh38
rs758660406 Chr.12: 101753389-101753389 on Build GRCh38
rs756642388 Chr.12: 101753493-101753493 on Build GRCh38
rs1005622047 Chr.12: 101753530-101753530 on Build GRCh38
rs781645250 Chr.12: 101753478-101753478 on Build GRCh38
rs281865039 Chr.12: 101753407-101753408 on Build GRCh38
rs576079970 Chr.12: 101753515-101753515 on Build GRCh38
rs764570097 Chr.12: 101753463-101753463 on Build GRCh38
rs201592854 Chr.12: 101753458-101753458 on Build GRCh38
rs182600512 Chr.12: 101753419-101753419 on Build GRCh38
rs748763888 Chr.12: 101753542-101753542 on Build GRCh38
rs144851964 Chr.12: 101753504-101753504 on Build GRCh38
rs281865037 Chr.12: 101753540-101753540 on Build GRCh38
rs925741743 Chr.12: 101753412-101753412 on Build GRCh38
rs766408752 Chr.12: 101753377-101753377 on Build GRCh38
rs281865020 Chr.12: 101753484-101753487 on Build GRCh38
rs753240668 Chr.12: 101753485-101753485 on Build GRCh38
rs777292254 Chr.12: 101753498-101753498 on Build GRCh38
rs281865018 Chr.12: 101753528-101753531 on Build GRCh38
rs769340984 Chr.12: 101753422-101753422 on Build GRCh38
rs979933501 Chr.12: 101753388-101753388 on Build GRCh38
rs187120606 Chr.12: 101753362-101753362 on Build GRCh38
rs281865019 Chr.12: 101753516-101753516 on Build GRCh38
rs79493678 Chr.12: 101753364-101753364 on Build GRCh38
rs774048511 Chr.12: 101753357-101753357 on Build GRCh38
rs281865038 Chr.12: 101753499-101753500 on Build GRCh38
rs780415252 Chr.12: 101753401-101753401 on Build GRCh38
rs376398528 Chr.12: 101753402-101753402 on Build GRCh38
rs149859473 Chr.12: 101753459-101753459 on Build GRCh38
rs1040259737 Chr.12: 101753445-101753445 on Build GRCh38
rs748934248 Chr.12: 101753440-101753440 on Build GRCh38
rs772131427 Chr.12: 101753449-101753449 on Build GRCh38
rs781689303 Chr.12: 101753413-101753414 on Build GRCh38
rs281865021 Chr.12: 101753445-101753451 on Build GRCh38

View Full Product Details