Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113011194-113011429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEPRO
Entrez Gene ID
25871
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
C3orf17, NET17
UniGene
Hs.591288
Gene Chromosome Location
Chr.3: 113002444-113019733 on Build GRCh38
Transcripts
NM_001319109.1 NM_001319110.1 NM_001319111.1 NM_001319112.1 NM_001319114.1 NM_001319115.1 NM_015412.3 NR_134969.1 XM_005247276.2 XM_011512635.2 XM_011512636.2 XM_011512638.2 XM_017006102.1 XM_017006103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5355138 c.758A>C p.K253T Chr.3: 113011200-113011200 on Build GRCh38
5451688 c.696G>A p.Q232Q Chr.3: 113011262-113011262 on Build GRCh38
4800808 c.595G>A p.E199K Chr.3: 113011363-113011363 on Build GRCh38
4112253 c.602T>A p.L201* Chr.3: 113011356-113011356 on Build GRCh38
6361674 c.709G>T p.A237S Chr.3: 113011249-113011249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776580705 Chr.3: 113011378-113011378 on Build GRCh38
rs747758567 Chr.3: 113011332-113011332 on Build GRCh38
rs766250023 Chr.3: 113011227-113011227 on Build GRCh38
rs765027824 Chr.3: 113011248-113011248 on Build GRCh38
rs908988822 Chr.3: 113011364-113011365 on Build GRCh38
rs769215170 Chr.3: 113011321-113011321 on Build GRCh38
rs746066669 Chr.3: 113011274-113011274 on Build GRCh38
rs759339702 Chr.3: 113011379-113011379 on Build GRCh38
rs763919380 Chr.3: 113011288-113011292 on Build GRCh38
rs780427753 Chr.3: 113011285-113011285 on Build GRCh38
rs771463764 Chr.3: 113011334-113011334 on Build GRCh38
rs896765182 Chr.3: 113011240-113011240 on Build GRCh38
rs572825067 Chr.3: 113011357-113011357 on Build GRCh38
rs781532668 Chr.3: 113011266-113011266 on Build GRCh38
rs979134867 Chr.3: 113011340-113011340 on Build GRCh38
rs757281270 Chr.3: 113011265-113011265 on Build GRCh38
rs757046366 Chr.3: 113011381-113011381 on Build GRCh38
rs202119046 Chr.3: 113011339-113011339 on Build GRCh38
rs754989548 Chr.3: 113011238-113011238 on Build GRCh38
rs765287779 Chr.3: 113011390-113011390 on Build GRCh38
rs753874841 Chr.3: 113011371-113011377 on Build GRCh38
rs770663450 Chr.3: 113011367-113011367 on Build GRCh38
rs749601707 Chr.3: 113011313-113011313 on Build GRCh38
rs138650982 Chr.3: 113011359-113011359 on Build GRCh38
rs1021175317 Chr.3: 113011338-113011338 on Build GRCh38
rs113746192 Chr.3: 113011231-113011231 on Build GRCh38
rs368142289 Chr.3: 113011256-113011256 on Build GRCh38
rs779217877 Chr.3: 113011325-113011325 on Build GRCh38
rs756509621 Chr.3: 113011282-113011282 on Build GRCh38

View Full Product Details