Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94824942-94825169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_017864.3 NR_073444.1 NR_073445.1 XM_017013616.1 XM_017013617.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6181715 c.242T>G p.L81* Chr.8: 94825004-94825004 on Build GRCh38
214536 c.282_283insGACTTA p.W94_D95insDL Chr.8: 94825044-94825045 on Build GRCh38
3902208 c.182C>A p.P61H Chr.8: 94824944-94824944 on Build GRCh38
1458734 c.236G>A p.R79H Chr.8: 94824998-94824998 on Build GRCh38
4772013 c.206delT p.Q70fs*12 Chr.8: 94824968-94824968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111939293 Chr.8: 94825057-94825057 on Build GRCh38
rs554102315 Chr.8: 94825088-94825088 on Build GRCh38
rs536918843 Chr.8: 94825136-94825136 on Build GRCh38
rs376914681 Chr.8: 94825063-94825063 on Build GRCh38
rs892383879 Chr.8: 94825130-94825130 on Build GRCh38
rs750777414 Chr.8: 94824990-94824990 on Build GRCh38
rs141032302 Chr.8: 94825062-94825062 on Build GRCh38
rs760133903 Chr.8: 94825115-94825115 on Build GRCh38
rs1020001229 Chr.8: 94825033-94825033 on Build GRCh38
rs931824865 Chr.8: 94825111-94825111 on Build GRCh38
rs770719611 Chr.8: 94825081-94825081 on Build GRCh38
rs947831994 Chr.8: 94825034-94825034 on Build GRCh38
rs151102552 Chr.8: 94824972-94824972 on Build GRCh38
rs781314077 Chr.8: 94825056-94825056 on Build GRCh38
rs910049286 Chr.8: 94825020-94825020 on Build GRCh38
rs755220058 Chr.8: 94825040-94825040 on Build GRCh38
rs369063673 Chr.8: 94824971-94824971 on Build GRCh38
rs771672175 Chr.8: 94825100-94825100 on Build GRCh38
rs141366808 Chr.8: 94825128-94825128 on Build GRCh38
rs749027022 Chr.8: 94825077-94825077 on Build GRCh38
rs762257751 Chr.8: 94825111-94825130 on Build GRCh38
rs373125263 Chr.8: 94825011-94825011 on Build GRCh38
rs751807833 Chr.8: 94825017-94825017 on Build GRCh38
rs1003183644 Chr.8: 94825120-94825120 on Build GRCh38
rs745522934 Chr.8: 94825094-94825094 on Build GRCh38
rs758853637 Chr.8: 94824996-94824996 on Build GRCh38
rs868594493 Chr.8: 94825075-94825075 on Build GRCh38

View Full Product Details