Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73196842-73197071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5549063 c.620C>G p.A207G Chr.17: 73196959-73196959 on Build GRCh38
4828489 c.701G>A p.R234K Chr.17: 73197040-73197040 on Build GRCh38
3521585 c.715C>G p.Q239E Chr.17: 73197054-73197054 on Build GRCh38
4827741 c.577G>A p.E193K Chr.17: 73196916-73196916 on Build GRCh38
6855391 c.625G>A p.A209T Chr.17: 73196964-73196964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765149692 Chr.17: 73197022-73197022 on Build GRCh38
rs141340733 Chr.17: 73196914-73196914 on Build GRCh38
rs201963725 Chr.17: 73196994-73196994 on Build GRCh38
rs771287810 Chr.17: 73196966-73196966 on Build GRCh38
rs778080401 Chr.17: 73196876-73196876 on Build GRCh38
rs370728414 Chr.17: 73196884-73196884 on Build GRCh38
rs752862204 Chr.17: 73196948-73196948 on Build GRCh38
rs189383307 Chr.17: 73197009-73197009 on Build GRCh38
rs750361604 Chr.17: 73196916-73196916 on Build GRCh38
rs567533944 Chr.17: 73196967-73196967 on Build GRCh38
rs759017136 Chr.17: 73196890-73196890 on Build GRCh38
rs145130181 Chr.17: 73196915-73196915 on Build GRCh38
rs193248551 Chr.17: 73196904-73196904 on Build GRCh38
rs778029188 Chr.17: 73196964-73196964 on Build GRCh38
rs931997867 Chr.17: 73196970-73196970 on Build GRCh38
rs771513054 Chr.17: 73196899-73196899 on Build GRCh38
rs185655840 Chr.17: 73196993-73196993 on Build GRCh38
rs750285470 Chr.17: 73197040-73197040 on Build GRCh38
rs765689135 Chr.17: 73197002-73197002 on Build GRCh38
rs749597821 Chr.17: 73196965-73196965 on Build GRCh38
rs141474045 Chr.17: 73196952-73196952 on Build GRCh38
rs781113656 Chr.17: 73196927-73196927 on Build GRCh38
rs1051883522 Chr.17: 73196999-73196999 on Build GRCh38
rs761663013 Chr.17: 73197020-73197020 on Build GRCh38
rs755041289 Chr.17: 73196922-73196922 on Build GRCh38
rs772683926 Chr.17: 73196982-73196982 on Build GRCh38
rs959870815 Chr.17: 73196909-73196909 on Build GRCh38
rs949454130 Chr.17: 73196870-73196870 on Build GRCh38
rs190156602 Chr.17: 73196877-73196877 on Build GRCh38
rs776431225 Chr.17: 73197017-73197017 on Build GRCh38
rs753679095 Chr.17: 73196913-73196913 on Build GRCh38
rs779512890 Chr.17: 73196885-73196887 on Build GRCh38
rs969009165 Chr.17: 73196947-73196947 on Build GRCh38
rs759814678 Chr.17: 73196879-73196879 on Build GRCh38
rs763875849 Chr.17: 73196905-73196905 on Build GRCh38
rs988562156 Chr.17: 73196912-73196912 on Build GRCh38

View Full Product Details