Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57563801-57564038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG5
Entrez Gene ID
221188
Gene Name
adhesion G protein-coupled receptor G5
Gene Aliases
GPR114, PGR27
UniGene
Hs.187884
Gene Chromosome Location
Chr.16: 57529339-57578182 on Build GRCh38
Transcripts
NM_001304376.2 NM_001318481.1 NM_153837.3 XM_011522949.2 XM_011522950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208593 c.361G>A p.A121T Chr.16: 57563911-57563911 on Build GRCh38
6673506 c.377C>A p.P126H Chr.16: 57563927-57563927 on Build GRCh38
4571003 c.364T>A p.C122S Chr.16: 57563914-57563914 on Build GRCh38
3402386 c.355C>T p.R119W Chr.16: 57563905-57563905 on Build GRCh38
6144732 c.369G>T p.K123N Chr.16: 57563919-57563919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759778232 Chr.16: 57563913-57563913 on Build GRCh38
rs536655478 Chr.16: 57563938-57563938 on Build GRCh38
rs201896883 Chr.16: 57563993-57563993 on Build GRCh38
rs753913510 Chr.16: 57563823-57563823 on Build GRCh38
rs760358667 Chr.16: 57563822-57563822 on Build GRCh38
rs368531040 Chr.16: 57563937-57563937 on Build GRCh38
rs754979215 Chr.16: 57563877-57563877 on Build GRCh38
rs372544241 Chr.16: 57563943-57563943 on Build GRCh38
rs200552943 Chr.16: 57563951-57563951 on Build GRCh38
rs375865051 Chr.16: 57563994-57563994 on Build GRCh38
rs944357303 Chr.16: 57564007-57564007 on Build GRCh38
rs367606004 Chr.16: 57563923-57563923 on Build GRCh38
rs773001740 Chr.16: 57563896-57563896 on Build GRCh38
rs766673893 Chr.16: 57563836-57563836 on Build GRCh38
rs778507357 Chr.16: 57563945-57563945 on Build GRCh38
rs752118180 Chr.16: 57564015-57564015 on Build GRCh38
rs746218229 Chr.16: 57563939-57563939 on Build GRCh38
rs755142904 Chr.16: 57563854-57563854 on Build GRCh38
rs547763306 Chr.16: 57563906-57563906 on Build GRCh38
rs772039795 Chr.16: 57563895-57563895 on Build GRCh38
rs931315350 Chr.16: 57563996-57563996 on Build GRCh38
rs576145241 Chr.16: 57563986-57563986 on Build GRCh38
rs150062848 Chr.16: 57563927-57563927 on Build GRCh38
rs765548451 Chr.16: 57563915-57563915 on Build GRCh38
rs139389524 Chr.16: 57563848-57563848 on Build GRCh38
rs373132751 Chr.16: 57563867-57563867 on Build GRCh38
rs371259029 Chr.16: 57563924-57563924 on Build GRCh38
rs35837636 Chr.16: 57563970-57563971 on Build GRCh38
rs747653288 Chr.16: 57563893-57563893 on Build GRCh38
rs984539239 Chr.16: 57564008-57564008 on Build GRCh38
rs753273804 Chr.16: 57563861-57563861 on Build GRCh38
rs375561538 Chr.16: 57564004-57564004 on Build GRCh38
rs764011691 Chr.16: 57564014-57564014 on Build GRCh38
rs200082143 Chr.16: 57563995-57563995 on Build GRCh38
rs771149116 Chr.16: 57563871-57563871 on Build GRCh38
rs748823110 Chr.16: 57563885-57563885 on Build GRCh38
rs746858485 Chr.16: 57563975-57563975 on Build GRCh38
rs759036606 Chr.16: 57563866-57563866 on Build GRCh38
rs546977099 Chr.16: 57563905-57563905 on Build GRCh38
rs779097441 Chr.16: 57563880-57563880 on Build GRCh38
rs779073168 Chr.16: 57563856-57563856 on Build GRCh38
rs763152708 Chr.16: 57564005-57564005 on Build GRCh38
rs377219588 Chr.16: 57563910-57563910 on Build GRCh38
rs768167624 Chr.16: 57563892-57563892 on Build GRCh38
rs201176226 Chr.16: 57563984-57563984 on Build GRCh38
rs773236823 Chr.16: 57563965-57563965 on Build GRCh38
rs747707971 Chr.16: 57563950-57563950 on Build GRCh38

View Full Product Details