Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200899402-200899651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf106
Entrez Gene ID
55765
Gene Name
chromosome 1 open reading frame 106
Gene Aliases
-
UniGene
Hs.518997
Gene Chromosome Location
Chr.1: 200891074-200915736 on Build GRCh38
Transcripts
NM_001142569.2 NM_018265.3 XM_011509754.2 XM_011509755.1 XM_011509756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4135902 c.378G>T p.L126L Chr.1: 200899540-200899540 on Build GRCh38
4135903 c.379G>T p.E127* Chr.1: 200899541-200899541 on Build GRCh38
6060602 c.323C>T p.P108L Chr.1: 200899485-200899485 on Build GRCh38
4804316 c.358C>T p.H120Y Chr.1: 200899520-200899520 on Build GRCh38
3481311 c.402G>A p.L134L Chr.1: 200899564-200899564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952977476 Chr.1: 200899474-200899474 on Build GRCh38
rs983060383 Chr.1: 200899462-200899462 on Build GRCh38
rs895685728 Chr.1: 200899508-200899508 on Build GRCh38
rs775794011 Chr.1: 200899569-200899569 on Build GRCh38
rs376201031 Chr.1: 200899460-200899460 on Build GRCh38
rs762007926 Chr.1: 200899586-200899586 on Build GRCh38
rs752643678 Chr.1: 200899606-200899606 on Build GRCh38
rs779553153 Chr.1: 200899613-200899613 on Build GRCh38
rs746460187 Chr.1: 200899454-200899454 on Build GRCh38
rs760768222 Chr.1: 200899596-200899596 on Build GRCh38
rs752447635 Chr.1: 200899501-200899501 on Build GRCh38
rs375903489 Chr.1: 200899610-200899610 on Build GRCh38
rs758120978 Chr.1: 200899425-200899425 on Build GRCh38
rs777425760 Chr.1: 200899523-200899523 on Build GRCh38
rs780740933 Chr.1: 200899557-200899557 on Build GRCh38
rs552128674 Chr.1: 200899589-200899589 on Build GRCh38
rs764996505 Chr.1: 200899500-200899500 on Build GRCh38
rs776043601 Chr.1: 200899485-200899485 on Build GRCh38
rs867224605 Chr.1: 200899597-200899597 on Build GRCh38
rs781010913 Chr.1: 200899628-200899628 on Build GRCh38
rs749602492 Chr.1: 200899568-200899568 on Build GRCh38
rs765090665 Chr.1: 200899602-200899602 on Build GRCh38
rs777320156 Chr.1: 200899444-200899444 on Build GRCh38
rs774445075 Chr.1: 200899491-200899491 on Build GRCh38
rs899542235 Chr.1: 200899590-200899590 on Build GRCh38
rs549475252 Chr.1: 200899478-200899478 on Build GRCh38
rs756905011 Chr.1: 200899627-200899627 on Build GRCh38
rs770360329 Chr.1: 200899456-200899456 on Build GRCh38
rs767551411 Chr.1: 200899495-200899495 on Build GRCh38
rs770248702 Chr.1: 200899477-200899477 on Build GRCh38
rs915109740 Chr.1: 200899616-200899616 on Build GRCh38
rs758130795 Chr.1: 200899515-200899515 on Build GRCh38
rs749910749 Chr.1: 200899629-200899629 on Build GRCh38
rs761898777 Chr.1: 200899492-200899492 on Build GRCh38
rs201328970 Chr.1: 200899546-200899546 on Build GRCh38
rs777790909 Chr.1: 200899455-200899456 on Build GRCh38
rs1041641805 Chr.1: 200899550-200899550 on Build GRCh38
rs751188115 Chr.1: 200899619-200899619 on Build GRCh38
rs1055628451 Chr.1: 200899505-200899505 on Build GRCh38
rs753770218 Chr.1: 200899496-200899496 on Build GRCh38
rs759511243 Chr.1: 200899497-200899497 on Build GRCh38
rs751227507 Chr.1: 200899532-200899532 on Build GRCh38
rs181832255 Chr.1: 200899545-200899545 on Build GRCh38
rs773330158 Chr.1: 200899595-200899595 on Build GRCh38
rs146632523 Chr.1: 200899486-200899486 on Build GRCh38
rs763862093 Chr.1: 200899617-200899617 on Build GRCh38
rs749831484 Chr.1: 200899540-200899540 on Build GRCh38
rs376986809 Chr.1: 200899489-200899489 on Build GRCh38

View Full Product Details