Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6862009-6862280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0232
Entrez Gene ID
9778
Gene Name
KIAA0232
Gene Aliases
-
UniGene
Hs.79276
Gene Chromosome Location
Chr.4: 6782732-6884172 on Build GRCh38
Transcripts
NM_001100590.1 NM_014743.2 XM_005248041.4 XM_011513598.2 XM_017008860.1 XM_017008861.1 XM_017008862.1 XM_017008863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
245208 c.1644G>T p.E548D Chr.4: 6862026-6862026 on Build GRCh38
1486052 c.1659A>T p.I553I Chr.4: 6862041-6862041 on Build GRCh38
1486053 c.1796G>C p.G599A Chr.4: 6862178-6862178 on Build GRCh38
3696709 c.1851C>G p.I617M Chr.4: 6862233-6862233 on Build GRCh38
4531366 c.1781G>C p.C594S Chr.4: 6862163-6862163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368542230 Chr.4: 6862176-6862176 on Build GRCh38
rs758972656 Chr.4: 6862258-6862258 on Build GRCh38
rs952838377 Chr.4: 6862221-6862221 on Build GRCh38
rs765759315 Chr.4: 6862091-6862091 on Build GRCh38
rs541080497 Chr.4: 6862229-6862229 on Build GRCh38
rs1034370736 Chr.4: 6862161-6862161 on Build GRCh38
rs776940791 Chr.4: 6862050-6862050 on Build GRCh38
rs781178164 Chr.4: 6862203-6862203 on Build GRCh38
rs200418556 Chr.4: 6862072-6862072 on Build GRCh38
rs199735011 Chr.4: 6862163-6862163 on Build GRCh38
rs765012260 Chr.4: 6862098-6862098 on Build GRCh38
rs759625084 Chr.4: 6862055-6862055 on Build GRCh38
rs201544484 Chr.4: 6862119-6862119 on Build GRCh38
rs780286719 Chr.4: 6862216-6862216 on Build GRCh38
rs752327429 Chr.4: 6862172-6862172 on Build GRCh38
rs757211576 Chr.4: 6862199-6862199 on Build GRCh38
rs753368742 Chr.4: 6862092-6862092 on Build GRCh38
rs773267441 Chr.4: 6862039-6862039 on Build GRCh38
rs570448167 Chr.4: 6862206-6862206 on Build GRCh38
rs772511469 Chr.4: 6862246-6862246 on Build GRCh38
rs774739483 Chr.4: 6862228-6862228 on Build GRCh38
rs762277529 Chr.4: 6862136-6862136 on Build GRCh38
rs775902799 Chr.4: 6862069-6862069 on Build GRCh38
rs199745129 Chr.4: 6862139-6862139 on Build GRCh38
rs371104885 Chr.4: 6862177-6862177 on Build GRCh38
rs370637729 Chr.4: 6862073-6862073 on Build GRCh38
rs754626245 Chr.4: 6862093-6862093 on Build GRCh38
rs375341830 Chr.4: 6862101-6862101 on Build GRCh38
rs773709829 Chr.4: 6862253-6862253 on Build GRCh38
rs755681713 Chr.4: 6862080-6862080 on Build GRCh38
rs748014042 Chr.4: 6862095-6862095 on Build GRCh38
rs764402749 Chr.4: 6862125-6862125 on Build GRCh38
rs745959761 Chr.4: 6862116-6862116 on Build GRCh38
rs556402927 Chr.4: 6862094-6862094 on Build GRCh38
rs763312088 Chr.4: 6862120-6862120 on Build GRCh38
rs758209918 Chr.4: 6862096-6862096 on Build GRCh38
rs765469831 Chr.4: 6862063-6862063 on Build GRCh38
rs774862274 Chr.4: 6862134-6862134 on Build GRCh38
rs769849682 Chr.4: 6862215-6862215 on Build GRCh38
rs751343465 Chr.4: 6862188-6862188 on Build GRCh38
rs753354028 Chr.4: 6862144-6862144 on Build GRCh38
rs367823638 Chr.4: 6862047-6862047 on Build GRCh38
rs368241178 Chr.4: 6862102-6862102 on Build GRCh38
rs939729619 Chr.4: 6862230-6862230 on Build GRCh38
rs962579375 Chr.4: 6862034-6862034 on Build GRCh38
rs201740172 Chr.4: 6862207-6862207 on Build GRCh38
rs766271910 Chr.4: 6862167-6862167 on Build GRCh38
rs749464651 Chr.4: 6862220-6862220 on Build GRCh38
rs372918347 Chr.4: 6862223-6862223 on Build GRCh38

View Full Product Details