Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107182338-107182563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBP1
Entrez Gene ID
26959
Gene Name
HMG-box transcription factor 1
Gene Aliases
-
UniGene
Hs.162032
Gene Chromosome Location
Chr.7: 107168961-107202529 on Build GRCh38
Transcripts
NM_001244262.1 NM_012257.3 XM_005250266.2 XM_005250267.2 XM_017011967.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5825289 c.213A>G p.Q71Q Chr.7: 107182416-107182416 on Build GRCh38
1083836 c.319G>T p.E107* Chr.7: 107182522-107182522 on Build GRCh38
5635689 c.231G>C p.L77L Chr.7: 107182434-107182434 on Build GRCh38
7370696 c.215C>G p.S72C Chr.7: 107182418-107182418 on Build GRCh38
6352752 c.346A>G p.N116D Chr.7: 107182549-107182549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949778025 Chr.7: 107182453-107182453 on Build GRCh38
rs756125534 Chr.7: 107182436-107182436 on Build GRCh38
rs778739583 Chr.7: 107182462-107182462 on Build GRCh38
rs999852061 Chr.7: 107182500-107182500 on Build GRCh38
rs763483953 Chr.7: 107182513-107182513 on Build GRCh38
rs763189955 Chr.7: 107182395-107182395 on Build GRCh38
rs766649119 Chr.7: 107182408-107182408 on Build GRCh38
rs1014992659 Chr.7: 107182394-107182394 on Build GRCh38
rs753687785 Chr.7: 107182446-107182446 on Build GRCh38
rs773545725 Chr.7: 107182512-107182512 on Build GRCh38
rs1005359743 Chr.7: 107182393-107182393 on Build GRCh38
rs773631987 Chr.7: 107182390-107182390 on Build GRCh38
rs759847372 Chr.7: 107182528-107182528 on Build GRCh38
rs752766934 Chr.7: 107182433-107182433 on Build GRCh38
rs774980601 Chr.7: 107182373-107182373 on Build GRCh38
rs973413546 Chr.7: 107182461-107182461 on Build GRCh38
rs745587590 Chr.7: 107182465-107182465 on Build GRCh38
rs368326211 Chr.7: 107182455-107182455 on Build GRCh38
rs779557957 Chr.7: 107182472-107182472 on Build GRCh38
rs568732709 Chr.7: 107182423-107182423 on Build GRCh38
rs767781110 Chr.7: 107182534-107182534 on Build GRCh38
rs774839245 Chr.7: 107182514-107182514 on Build GRCh38
rs963339708 Chr.7: 107182415-107182415 on Build GRCh38
rs201453687 Chr.7: 107182470-107182470 on Build GRCh38
rs746453456 Chr.7: 107182487-107182487 on Build GRCh38
rs767760471 Chr.7: 107182424-107182424 on Build GRCh38
rs980054679 Chr.7: 107182496-107182496 on Build GRCh38
rs143622240 Chr.7: 107182493-107182493 on Build GRCh38
rs139991246 Chr.7: 107182380-107182380 on Build GRCh38
rs751867690 Chr.7: 107182416-107182416 on Build GRCh38
rs777530166 Chr.7: 107182442-107182442 on Build GRCh38
rs376024723 Chr.7: 107182378-107182378 on Build GRCh38
rs750790118 Chr.7: 107182387-107182387 on Build GRCh38

View Full Product Details