Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 131003146-131003379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRD1
Entrez Gene ID
283383
Gene Name
adhesion G protein-coupled receptor D1
Gene Aliases
GPR133, PGR25
UniGene
Hs.719239
Gene Chromosome Location
Chr.12: 130953907-131141469 on Build GRCh38
Transcripts
NM_198827.3 XM_005253566.2 XM_011538203.2 XM_011538204.2 XM_011538205.2 XM_011538206.2 XM_011538207.2 XM_011538208.2 XM_011538209.2 XM_011538210.2 XM_011538211.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459079 c.1110C>T p.P370P Chr.12: 131003268-131003268 on Build GRCh38
3458151 c.1137C>T p.A379A Chr.12: 131003295-131003295 on Build GRCh38
5704349 c.1055A>G p.D352G Chr.12: 131003213-131003213 on Build GRCh38
7158292 c.1038G>A p.V346V Chr.12: 131003196-131003196 on Build GRCh38
1677481 c.1082T>C p.V361A Chr.12: 131003240-131003240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776616695 Chr.12: 131003246-131003246 on Build GRCh38
rs142314859 Chr.12: 131003264-131003264 on Build GRCh38
rs572513767 Chr.12: 131003191-131003191 on Build GRCh38
rs781621386 Chr.12: 131003211-131003211 on Build GRCh38
rs756490150 Chr.12: 131003217-131003217 on Build GRCh38
rs140194684 Chr.12: 131003329-131003329 on Build GRCh38
rs773324312 Chr.12: 131003178-131003178 on Build GRCh38
rs769599446 Chr.12: 131003250-131003250 on Build GRCh38
rs758138615 Chr.12: 131003354-131003354 on Build GRCh38
rs563561024 Chr.12: 131003316-131003316 on Build GRCh38
rs746975895 Chr.12: 131003237-131003237 on Build GRCh38
rs778501649 Chr.12: 131003218-131003218 on Build GRCh38
rs532956146 Chr.12: 131003265-131003265 on Build GRCh38
rs146260772 Chr.12: 131003190-131003190 on Build GRCh38
rs368192987 Chr.12: 131003342-131003342 on Build GRCh38
rs758512461 Chr.12: 131003193-131003193 on Build GRCh38
rs950118954 Chr.12: 131003297-131003297 on Build GRCh38
rs776250054 Chr.12: 131003188-131003188 on Build GRCh38
rs375414030 Chr.12: 131003226-131003226 on Build GRCh38
rs373680341 Chr.12: 131003341-131003341 on Build GRCh38
rs1046276448 Chr.12: 131003313-131003313 on Build GRCh38
rs750699249 Chr.12: 131003214-131003214 on Build GRCh38
rs146055067 Chr.12: 131003229-131003229 on Build GRCh38
rs549833008 Chr.12: 131003277-131003277 on Build GRCh38
rs775221745 Chr.12: 131003321-131003321 on Build GRCh38
rs781031815 Chr.12: 131003291-131003291 on Build GRCh38
rs200670259 Chr.12: 131003181-131003181 on Build GRCh38
rs148751019 Chr.12: 131003256-131003256 on Build GRCh38
rs770468941 Chr.12: 131003288-131003288 on Build GRCh38
rs764133268 Chr.12: 131003194-131003194 on Build GRCh38
rs549029202 Chr.12: 131003339-131003339 on Build GRCh38
rs565822971 Chr.12: 131003344-131003344 on Build GRCh38
rs745794956 Chr.12: 131003312-131003312 on Build GRCh38
rs747288448 Chr.12: 131003278-131003278 on Build GRCh38
rs370854685 Chr.12: 131003204-131003204 on Build GRCh38
rs780445686 Chr.12: 131003267-131003267 on Build GRCh38
rs770707816 Chr.12: 131003238-131003238 on Build GRCh38
rs146050435 Chr.12: 131003249-131003249 on Build GRCh38
rs768805668 Chr.12: 131003332-131003332 on Build GRCh38
rs766881728 Chr.12: 131003345-131003345 on Build GRCh38
rs746574998 Chr.12: 131003286-131003286 on Build GRCh38
rs745315105 Chr.12: 131003239-131003240 on Build GRCh38
rs762399199 Chr.12: 131003261-131003261 on Build GRCh38
rs763109065 Chr.12: 131003253-131003253 on Build GRCh38
rs754191350 Chr.12: 131003346-131003346 on Build GRCh38

View Full Product Details