Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197938583-197938844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCG
Entrez Gene ID
84223
Gene Name
IQ motif containing G
Gene Aliases
CFAP122, DRC9
UniGene
Hs.591675
Gene Chromosome Location
Chr.3: 197889075-197960019 on Build GRCh38
Transcripts
NM_001134435.2 NM_001323027.1 NM_001323028.1 NM_001323029.1 NM_001323030.1 NM_032263.4 XM_017007316.1 XM_017007317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002624 c.335C>A p.A112D Chr.3: 197938728-197938728 on Build GRCh38
6358259 c.418G>A p.A140T Chr.3: 197938645-197938645 on Build GRCh38
1308997 c.373G>A p.E125K Chr.3: 197938690-197938690 on Build GRCh38
4584229 c.388T>C p.L130L Chr.3: 197938675-197938675 on Build GRCh38
4002623 c.335C>G p.A112G Chr.3: 197938728-197938728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775606625 Chr.3: 197938788-197938788 on Build GRCh38
rs9880989 Chr.3: 197938728-197938728 on Build GRCh38
rs549040318 Chr.3: 197938675-197938675 on Build GRCh38
rs781318400 Chr.3: 197938753-197938753 on Build GRCh38
rs200494755 Chr.3: 197938809-197938810 on Build GRCh38
rs75671851 Chr.3: 197938650-197938650 on Build GRCh38
rs201676602 Chr.3: 197938708-197938708 on Build GRCh38
rs377674167 Chr.3: 197938723-197938723 on Build GRCh38
rs202067997 Chr.3: 197938620-197938620 on Build GRCh38
rs371193619 Chr.3: 197938807-197938807 on Build GRCh38
rs766841659 Chr.3: 197938646-197938646 on Build GRCh38
rs760723084 Chr.3: 197938793-197938793 on Build GRCh38
rs141469670 Chr.3: 197938748-197938748 on Build GRCh38
rs767907505 Chr.3: 197938785-197938785 on Build GRCh38
rs753684079 Chr.3: 197938688-197938688 on Build GRCh38
rs749706380 Chr.3: 197938771-197938771 on Build GRCh38
rs762363015 Chr.3: 197938654-197938655 on Build GRCh38
rs143910661 Chr.3: 197938751-197938751 on Build GRCh38
rs746511531 Chr.3: 197938738-197938738 on Build GRCh38
rs760024667 Chr.3: 197938784-197938784 on Build GRCh38
rs145406608 Chr.3: 197938747-197938747 on Build GRCh38
rs774738135 Chr.3: 197938783-197938783 on Build GRCh38
rs780354023 Chr.3: 197938711-197938711 on Build GRCh38
rs764341379 Chr.3: 197938795-197938795 on Build GRCh38
rs771282398 Chr.3: 197938779-197938779 on Build GRCh38
rs139671579 Chr.3: 197938651-197938651 on Build GRCh38
rs770285576 Chr.3: 197938633-197938633 on Build GRCh38
rs372696867 Chr.3: 197938715-197938715 on Build GRCh38
rs773778015 Chr.3: 197938634-197938634 on Build GRCh38
rs149674426 Chr.3: 197938732-197938732 on Build GRCh38
rs765276819 Chr.3: 197938802-197938805 on Build GRCh38
rs369565790 Chr.3: 197938764-197938764 on Build GRCh38
rs150540903 Chr.3: 197938683-197938683 on Build GRCh38
rs745768328 Chr.3: 197938727-197938727 on Build GRCh38
rs202218844 Chr.3: 197938672-197938672 on Build GRCh38
rs765024172 Chr.3: 197938808-197938808 on Build GRCh38
rs542436769 Chr.3: 197938766-197938766 on Build GRCh38
rs771570790 Chr.3: 197938619-197938619 on Build GRCh38
rs750330845 Chr.3: 197938809-197938809 on Build GRCh38
rs760344275 Chr.3: 197938632-197938632 on Build GRCh38
rs142802587 Chr.3: 197938645-197938645 on Build GRCh38
rs761509940 Chr.3: 197938644-197938644 on Build GRCh38

View Full Product Details