Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113942999-113943230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR3B
Entrez Gene ID
9177
Gene Name
5-hydroxytryptamine receptor 3B
Gene Aliases
5-HT3B
UniGene
Hs.241377
Gene Chromosome Location
Chr.11: 113904796-113949682 on Build GRCh38
Transcripts
NM_006028.4 XM_011543063.1 XM_017018552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6131359 c.793C>T p.P265S Chr.11: 113943078-113943078 on Build GRCh38
5323449 c.848G>A p.R283Q Chr.11: 113943166-113943166 on Build GRCh38
6131360 c.760C>T p.P254S Chr.11: 113943078-113943078 on Build GRCh38
2155372 c.741G>C p.L247L Chr.11: 113943026-113943026 on Build GRCh38
7194460 c.848T>C p.V283A Chr.11: 113943133-113943133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758245312 Chr.11: 113943019-113943019 on Build GRCh38
rs142289832 Chr.11: 113943163-113943163 on Build GRCh38
rs768426203 Chr.11: 113943034-113943034 on Build GRCh38
rs767225320 Chr.11: 113943204-113943205 on Build GRCh38
rs549253438 Chr.11: 113943095-113943095 on Build GRCh38
rs770130259 Chr.11: 113943143-113943143 on Build GRCh38
rs779856472 Chr.11: 113943119-113943119 on Build GRCh38
rs201779669 Chr.11: 113943069-113943069 on Build GRCh38
rs375381760 Chr.11: 113943204-113943204 on Build GRCh38
rs539145522 Chr.11: 113943131-113943131 on Build GRCh38
rs148829936 Chr.11: 113943088-113943088 on Build GRCh38
rs749886032 Chr.11: 113943104-113943104 on Build GRCh38
rs145795113 Chr.11: 113943166-113943166 on Build GRCh38
rs544377745 Chr.11: 113943165-113943165 on Build GRCh38
rs760047207 Chr.11: 113943197-113943197 on Build GRCh38
rs139008859 Chr.11: 113943154-113943154 on Build GRCh38
rs372054196 Chr.11: 113943202-113943202 on Build GRCh38
rs781448059 Chr.11: 113943144-113943144 on Build GRCh38
rs777003399 Chr.11: 113943037-113943037 on Build GRCh38
rs758021625 Chr.11: 113943121-113943121 on Build GRCh38
rs756867912 Chr.11: 113943074-113943074 on Build GRCh38
rs908520742 Chr.11: 113943205-113943205 on Build GRCh38
rs138362802 Chr.11: 113943078-113943078 on Build GRCh38
rs970549409 Chr.11: 113943099-113943099 on Build GRCh38
rs770037424 Chr.11: 113943056-113943056 on Build GRCh38
rs746773022 Chr.11: 113943027-113943027 on Build GRCh38
rs781107106 Chr.11: 113943132-113943132 on Build GRCh38
rs945265292 Chr.11: 113943177-113943177 on Build GRCh38
rs779954110 Chr.11: 113943022-113943022 on Build GRCh38
rs372635147 Chr.11: 113943068-113943068 on Build GRCh38
rs751013344 Chr.11: 113943117-113943118 on Build GRCh38
rs765675345 Chr.11: 113943080-113943081 on Build GRCh38
rs748343438 Chr.11: 113943054-113943054 on Build GRCh38
rs746902797 Chr.11: 113943130-113943130 on Build GRCh38
rs145278314 Chr.11: 113943097-113943097 on Build GRCh38
rs981136949 Chr.11: 113943113-113943113 on Build GRCh38
rs766793685 Chr.11: 113943073-113943073 on Build GRCh38
rs542556393 Chr.11: 113943087-113943087 on Build GRCh38
rs1046885747 Chr.11: 113943193-113943193 on Build GRCh38
rs754473348 Chr.11: 113943179-113943180 on Build GRCh38
rs767821533 Chr.11: 113943080-113943080 on Build GRCh38

View Full Product Details