Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133956768-133957041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf15
Entrez Gene ID
56951
Gene Name
chromosome 5 open reading frame 15
Gene Aliases
HTGN29, KCT2
UniGene
Hs.744034
Gene Chromosome Location
Chr.5: 133955507-133968715 on Build GRCh38
Transcripts
NM_020199.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5729992 c.733C>T p.R245C Chr.5: 133956924-133956924 on Build GRCh38
1060834 c.704G>A p.G235D Chr.5: 133956953-133956953 on Build GRCh38
4667799 c.773A>T p.K258M Chr.5: 133956884-133956884 on Build GRCh38
3850117 c.732T>C p.H244H Chr.5: 133956925-133956925 on Build GRCh38
4667800 c.727T>C p.Y243H Chr.5: 133956930-133956930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187246788 Chr.5: 133956953-133956953 on Build GRCh38
rs746656501 Chr.5: 133956809-133956809 on Build GRCh38
rs541713224 Chr.5: 133956810-133956810 on Build GRCh38
rs753942566 Chr.5: 133956898-133956898 on Build GRCh38
rs750589127 Chr.5: 133956878-133956878 on Build GRCh38
rs762294013 Chr.5: 133956959-133956959 on Build GRCh38
rs775052686 Chr.5: 133956946-133956946 on Build GRCh38
rs766514661 Chr.5: 133956897-133956897 on Build GRCh38
rs199505921 Chr.5: 133957007-133957007 on Build GRCh38
rs139570849 Chr.5: 133956931-133956931 on Build GRCh38
rs752638440 Chr.5: 133956956-133956956 on Build GRCh38
rs146335440 Chr.5: 133956875-133956875 on Build GRCh38
rs61730680 Chr.5: 133956955-133956955 on Build GRCh38
rs767784325 Chr.5: 133956867-133956867 on Build GRCh38
rs374778255 Chr.5: 133956815-133956815 on Build GRCh38
rs372276960 Chr.5: 133956851-133956851 on Build GRCh38
rs746634804 Chr.5: 133956995-133956995 on Build GRCh38
rs755153221 Chr.5: 133956911-133956911 on Build GRCh38
rs923503504 Chr.5: 133956924-133956924 on Build GRCh38
rs760835895 Chr.5: 133956890-133956890 on Build GRCh38
rs776150380 Chr.5: 133956819-133956819 on Build GRCh38
rs76685384 Chr.5: 133956994-133956994 on Build GRCh38
rs964797466 Chr.5: 133956846-133956846 on Build GRCh38
rs774904834 Chr.5: 133956840-133956840 on Build GRCh38
rs368939447 Chr.5: 133956831-133956831 on Build GRCh38
rs933464035 Chr.5: 133956948-133956948 on Build GRCh38
rs778993410 Chr.5: 133956923-133956923 on Build GRCh38
rs777436502 Chr.5: 133956989-133956989 on Build GRCh38

View Full Product Details