Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22833307-22833574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL52
Entrez Gene ID
122704
Gene Name
mitochondrial ribosomal protein L52
Gene Aliases
-
UniGene
Hs.355935
Gene Chromosome Location
Chr.14: 22829860-22835037 on Build GRCh38
Transcripts
NM_001318870.1 NM_178336.2 NM_180982.2 NM_181304.2 NM_181305.2 NM_181306.2 NM_181307.2 XM_005267325.4 XM_005267328.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955910 c.209A>G p.E70G Chr.14: 22833472-22833472 on Build GRCh38
432855 c.195A>G p.R65R Chr.14: 22833458-22833458 on Build GRCh38
5971105 c.170C>T p.P57L Chr.14: 22833433-22833433 on Build GRCh38
7270568 c.217G>T p.A73S Chr.14: 22833480-22833480 on Build GRCh38
2030548 c.159C>T p.G53G Chr.14: 22833422-22833422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17122962 Chr.14: 22833356-22833356 on Build GRCh38
rs556782879 Chr.14: 22833370-22833370 on Build GRCh38
rs34799922 Chr.14: 22833550-22833551 on Build GRCh38
rs910600206 Chr.14: 22833409-22833409 on Build GRCh38
rs769888738 Chr.14: 22833528-22833528 on Build GRCh38
rs567192462 Chr.14: 22833403-22833404 on Build GRCh38
rs972112131 Chr.14: 22833336-22833336 on Build GRCh38
rs761852248 Chr.14: 22833502-22833502 on Build GRCh38
rs780510049 Chr.14: 22833482-22833482 on Build GRCh38
rs767300375 Chr.14: 22833536-22833536 on Build GRCh38
rs111564654 Chr.14: 22833416-22833416 on Build GRCh38
rs542832940 Chr.14: 22833454-22833454 on Build GRCh38
rs781099882 Chr.14: 22833487-22833490 on Build GRCh38
rs767249100 Chr.14: 22833429-22833429 on Build GRCh38
rs770493413 Chr.14: 22833527-22833527 on Build GRCh38
rs747523588 Chr.14: 22833486-22833486 on Build GRCh38
rs749138620 Chr.14: 22833496-22833497 on Build GRCh38
rs745578127 Chr.14: 22833350-22833350 on Build GRCh38
rs755934125 Chr.14: 22833431-22833431 on Build GRCh38
rs763100966 Chr.14: 22833534-22833534 on Build GRCh38
rs766814719 Chr.14: 22833475-22833475 on Build GRCh38
rs78923701 Chr.14: 22833434-22833434 on Build GRCh38
rs201749208 Chr.14: 22833438-22833438 on Build GRCh38
rs750625277 Chr.14: 22833423-22833423 on Build GRCh38
rs765439695 Chr.14: 22833418-22833418 on Build GRCh38
rs12050397 Chr.14: 22833376-22833376 on Build GRCh38
rs377615020 Chr.14: 22833367-22833367 on Build GRCh38
rs899650138 Chr.14: 22833371-22833371 on Build GRCh38
rs146260711 Chr.14: 22833476-22833476 on Build GRCh38
rs367761850 Chr.14: 22833500-22833500 on Build GRCh38
rs745475160 Chr.14: 22833465-22833465 on Build GRCh38
rs866072883 Chr.14: 22833439-22833439 on Build GRCh38
rs773456784 Chr.14: 22833530-22833530 on Build GRCh38
rs553925588 Chr.14: 22833505-22833506 on Build GRCh38
rs142727241 Chr.14: 22833497-22833497 on Build GRCh38
rs138366764 Chr.14: 22833424-22833424 on Build GRCh38
rs912478849 Chr.14: 22833353-22833353 on Build GRCh38
rs113354123 Chr.14: 22833415-22833415 on Build GRCh38
rs370351677 Chr.14: 22833453-22833453 on Build GRCh38

View Full Product Details