Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169394361-169394618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK1
Entrez Gene ID
4750
Gene Name
NIMA related kinase 1
Gene Aliases
NY-REN-55, SRPS2, SRPS2A, SRTD6
UniGene
Hs.481181
Gene Chromosome Location
Chr.4: 169393270-169612629 on Build GRCh38
Transcripts
NM_001199397.1 NM_001199398.1 NM_001199399.1 NM_001199400.1 NM_012224.2 XM_006714228.1 XM_011532003.1 XM_011532004.1 XM_017008249.1 XM_017008250.1 XM_017008251.1 XM_017008252.1 XM_017008253.1 XM_017008254.1

SNP Detail

SNP ID Chromosome Location
rs372935588 Chr.4: 169394491-169394491 on Build GRCh38
rs749195740 Chr.4: 169394487-169394487 on Build GRCh38
rs187803112 Chr.4: 169394450-169394450 on Build GRCh38
rs767808092 Chr.4: 169394403-169394403 on Build GRCh38
rs575318861 Chr.4: 169394556-169394556 on Build GRCh38
rs1057235350 Chr.4: 169394531-169394531 on Build GRCh38
rs756139766 Chr.4: 169394492-169394493 on Build GRCh38
rs76330049 Chr.4: 169394393-169394393 on Build GRCh38
rs776932483 Chr.4: 169394526-169394526 on Build GRCh38
rs753004145 Chr.4: 169394544-169394544 on Build GRCh38
rs917291288 Chr.4: 169394577-169394578 on Build GRCh38
rs908699767 Chr.4: 169394521-169394521 on Build GRCh38
rs1004584477 Chr.4: 169394412-169394412 on Build GRCh38
rs183545840 Chr.4: 169394400-169394400 on Build GRCh38
rs777864764 Chr.4: 169394493-169394493 on Build GRCh38
rs895882142 Chr.4: 169394457-169394457 on Build GRCh38
rs749160784 Chr.4: 169394535-169394536 on Build GRCh38
rs747841498 Chr.4: 169394473-169394473 on Build GRCh38
rs762040322 Chr.4: 169394533-169394533 on Build GRCh38
rs755925133 Chr.4: 169394483-169394483 on Build GRCh38
rs865877221 Chr.4: 169394402-169394402 on Build GRCh38
rs879165314 Chr.4: 169394566-169394566 on Build GRCh38
rs780097631 Chr.4: 169394505-169394505 on Build GRCh38
rs535517817 Chr.4: 169394484-169394484 on Build GRCh38
rs768700318 Chr.4: 169394519-169394519 on Build GRCh38

View Full Product Details