Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7190419-7190664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX5
Entrez Gene ID
5830
Gene Name
peroxisomal biogenesis factor 5
Gene Aliases
PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
UniGene
Hs.567327
Gene Chromosome Location
Chr.12: 7188072-7218574 on Build GRCh38
Transcripts
NM_000319.4 NM_001131023.1 NM_001131024.1 NM_001131025.1 NM_001131026.1 NM_001300789.1 XM_005253455.1 XM_011520793.2 XM_011520795.1 XM_011520797.1 XM_011520798.1 XM_011520799.2 XM_011520800.1 XM_011520801.2 XM_011520802.2 XM_017019744.1 XM_017019745.1 XM_017019746.1 XM_017019747.1 XM_017019748.1 XM_017019749.1 XM_017019750.1 XM_017019751.1 XM_017019752.1 XM_017019753.1 XM_017019754.1 XM_017019755.1 XM_017019756.1 XM_017019757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4044754 c.64G>A p.G22R Chr.12: 7190441-7190441 on Build GRCh38
3465143 c.88C>T p.L30F Chr.12: 7190465-7190465 on Build GRCh38
5201604 c.78G>A p.Q26Q Chr.12: 7190455-7190455 on Build GRCh38
6310322 c.135G>A p.P45P Chr.12: 7190512-7190512 on Build GRCh38
1220225 c.113G>A p.G38D Chr.12: 7190490-7190490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778434457 Chr.12: 7190515-7190557 on Build GRCh38
rs773639278 Chr.12: 7190568-7190568 on Build GRCh38
rs758936844 Chr.12: 7190479-7190479 on Build GRCh38
rs867279144 Chr.12: 7190601-7190601 on Build GRCh38
rs753239589 Chr.12: 7190476-7190476 on Build GRCh38
rs145649593 Chr.12: 7190511-7190511 on Build GRCh38
rs761815459 Chr.12: 7190565-7190608 on Build GRCh38
rs906121777 Chr.12: 7190639-7190639 on Build GRCh38
rs1014494503 Chr.12: 7190608-7190608 on Build GRCh38
rs770949712 Chr.12: 7190452-7190452 on Build GRCh38
rs869196302 Chr.12: 7190512-7190557 on Build GRCh38
rs756732117 Chr.12: 7190512-7190513 on Build GRCh38
rs749342175 Chr.12: 7190528-7190528 on Build GRCh38
rs751937990 Chr.12: 7190492-7190492 on Build GRCh38
rs766807646 Chr.12: 7190483-7190483 on Build GRCh38
rs746986459 Chr.12: 7190552-7190592 on Build GRCh38
rs777383921 Chr.12: 7190497-7190498 on Build GRCh38
rs932532585 Chr.12: 7190500-7190500 on Build GRCh38
rs1052760898 Chr.12: 7190563-7190563 on Build GRCh38
rs190526209 Chr.12: 7190469-7190469 on Build GRCh38
rs768505581 Chr.12: 7190558-7190563 on Build GRCh38
rs398123572 Chr.12: 7190458-7190458 on Build GRCh38
rs759345862 Chr.12: 7190460-7190460 on Build GRCh38
rs757612863 Chr.12: 7190513-7190557 on Build GRCh38
rs767406855 Chr.12: 7190468-7190468 on Build GRCh38
rs771908923 Chr.12: 7190536-7190579 on Build GRCh38
rs749267891 Chr.12: 7190443-7190443 on Build GRCh38
rs763255641 Chr.12: 7190570-7190613 on Build GRCh38
rs748309745 Chr.12: 7190438-7190438 on Build GRCh38
rs781410836 Chr.12: 7190498-7190498 on Build GRCh38
rs374590365 Chr.12: 7190440-7190440 on Build GRCh38
rs769432821 Chr.12: 7190470-7190470 on Build GRCh38
rs751943129 Chr.12: 7190594-7190628 on Build GRCh38
rs766900011 Chr.12: 7190582-7190591 on Build GRCh38
rs74528900 Chr.12: 7190628-7190628 on Build GRCh38
rs895521309 Chr.12: 7190580-7190580 on Build GRCh38
rs776586006 Chr.12: 7190563-7190563 on Build GRCh38
rs745513975 Chr.12: 7190534-7190534 on Build GRCh38
rs183090244 Chr.12: 7190583-7190583 on Build GRCh38
rs11269713 Chr.12: 7190645-7190646 on Build GRCh38
rs775238354 Chr.12: 7190547-7190550 on Build GRCh38
rs976676879 Chr.12: 7190446-7190446 on Build GRCh38
rs765298252 Chr.12: 7190566-7190604 on Build GRCh38
rs761885230 Chr.12: 7190512-7190512 on Build GRCh38
rs748769764 Chr.12: 7190507-7190514 on Build GRCh38
rs772786048 Chr.12: 7190499-7190499 on Build GRCh38
rs543472765 Chr.12: 7190517-7190517 on Build GRCh38
rs1135240 Chr.12: 7190509-7190509 on Build GRCh38
rs934412084 Chr.12: 7190535-7190535 on Build GRCh38
rs372992555 Chr.12: 7190508-7190508 on Build GRCh38
rs932238098 Chr.12: 7190506-7190506 on Build GRCh38

View Full Product Details