Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44254664-44254937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35B2
Entrez Gene ID
347734
Gene Name
solute carrier family 35 member B2
Gene Aliases
PAPST1, SLL, UGTrel4
UniGene
Hs.182885
Gene Chromosome Location
Chr.6: 44254101-44257890 on Build GRCh38
Transcripts
NM_001286509.1 NM_001286510.1 NM_001286511.1 NM_001286512.1 NM_001286513.1 NM_001286517.1 NM_001286519.1 NM_001286520.1 NM_178148.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842107 c.1252C>T p.R418W Chr.6: 44254753-44254753 on Build GRCh38
3394223 c.1235G>A p.R412Q Chr.6: 44254770-44254770 on Build GRCh38
4445060 c.1288C>T p.Q430* Chr.6: 44254717-44254717 on Build GRCh38
451551 c.1232C>T p.A411V Chr.6: 44254773-44254773 on Build GRCh38
6107013 c.1221C>G p.L407L Chr.6: 44254784-44254784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138499828 Chr.6: 44254753-44254753 on Build GRCh38
rs368956564 Chr.6: 44254712-44254712 on Build GRCh38
rs150122797 Chr.6: 44254729-44254729 on Build GRCh38
rs372657328 Chr.6: 44254772-44254772 on Build GRCh38
rs61740055 Chr.6: 44254774-44254774 on Build GRCh38
rs771790987 Chr.6: 44254726-44254726 on Build GRCh38
rs934034301 Chr.6: 44254725-44254725 on Build GRCh38
rs565377883 Chr.6: 44254744-44254744 on Build GRCh38
rs547053957 Chr.6: 44254727-44254727 on Build GRCh38
rs146186426 Chr.6: 44254775-44254775 on Build GRCh38
rs1029591429 Chr.6: 44254885-44254885 on Build GRCh38
rs748829958 Chr.6: 44254832-44254832 on Build GRCh38
rs769418624 Chr.6: 44254813-44254813 on Build GRCh38
rs768241877 Chr.6: 44254916-44254916 on Build GRCh38
rs963934077 Chr.6: 44254862-44254862 on Build GRCh38
rs201389938 Chr.6: 44254797-44254797 on Build GRCh38
rs200869630 Chr.6: 44254828-44254828 on Build GRCh38
rs200968592 Chr.6: 44254765-44254765 on Build GRCh38
rs528326020 Chr.6: 44254745-44254745 on Build GRCh38
rs780009082 Chr.6: 44254796-44254796 on Build GRCh38
rs764817649 Chr.6: 44254707-44254708 on Build GRCh38
rs975393659 Chr.6: 44254714-44254714 on Build GRCh38
rs1013942947 Chr.6: 44254777-44254777 on Build GRCh38
rs761110971 Chr.6: 44254778-44254778 on Build GRCh38
rs376787684 Chr.6: 44254914-44254914 on Build GRCh38
rs528603457 Chr.6: 44254721-44254721 on Build GRCh38
rs556702896 Chr.6: 44254837-44254837 on Build GRCh38
rs764671985 Chr.6: 44254734-44254734 on Build GRCh38
rs749351754 Chr.6: 44254709-44254709 on Build GRCh38
rs779091751 Chr.6: 44254757-44254757 on Build GRCh38
rs748049137 Chr.6: 44254724-44254724 on Build GRCh38
rs758518037 Chr.6: 44254880-44254880 on Build GRCh38
rs748054554 Chr.6: 44254759-44254759 on Build GRCh38
rs759032589 Chr.6: 44254730-44254730 on Build GRCh38
rs551150308 Chr.6: 44254752-44254752 on Build GRCh38
rs760193934 Chr.6: 44254855-44254855 on Build GRCh38
rs763729364 Chr.6: 44254758-44254759 on Build GRCh38
rs751441824 Chr.6: 44254760-44254761 on Build GRCh38
rs200194380 Chr.6: 44254767-44254767 on Build GRCh38
rs202082767 Chr.6: 44254799-44254799 on Build GRCh38
rs757813848 Chr.6: 44254742-44254744 on Build GRCh38
rs141587728 Chr.6: 44254768-44254768 on Build GRCh38
rs35163667 Chr.6: 44254847-44254847 on Build GRCh38
rs755794449 Chr.6: 44254909-44254909 on Build GRCh38
rs770539467 Chr.6: 44254728-44254728 on Build GRCh38
rs780334263 Chr.6: 44254705-44254705 on Build GRCh38
rs748837432 Chr.6: 44254910-44254910 on Build GRCh38
rs762279662 Chr.6: 44254773-44254773 on Build GRCh38
rs781554904 Chr.6: 44254694-44254694 on Build GRCh38
rs766780517 Chr.6: 44254782-44254782 on Build GRCh38
rs776004233 Chr.6: 44254871-44254871 on Build GRCh38
rs752013909 Chr.6: 44254736-44254736 on Build GRCh38
rs886864739 Chr.6: 44254831-44254831 on Build GRCh38
rs757164153 Chr.6: 44254780-44254781 on Build GRCh38
rs1059894 Chr.6: 44254762-44254762 on Build GRCh38
rs752775711 Chr.6: 44254879-44254879 on Build GRCh38
rs375941077 Chr.6: 44254810-44254810 on Build GRCh38
rs778904736 Chr.6: 44254793-44254793 on Build GRCh38
rs772022635 Chr.6: 44254764-44254764 on Build GRCh38
rs775017717 Chr.6: 44254771-44254771 on Build GRCh38
rs765492376 Chr.6: 44254868-44254868 on Build GRCh38
rs755110539 Chr.6: 44254754-44254754 on Build GRCh38
rs760851221 Chr.6: 44254785-44254787 on Build GRCh38
rs748462914 Chr.6: 44254742-44254742 on Build GRCh38
rs941263698 Chr.6: 44254808-44254808 on Build GRCh38
rs375605580 Chr.6: 44254704-44254704 on Build GRCh38
rs375633304 Chr.6: 44254700-44254700 on Build GRCh38
rs532752079 Chr.6: 44254748-44254748 on Build GRCh38
rs777110019 Chr.6: 44254859-44254859 on Build GRCh38
rs752392556 Chr.6: 44254712-44254714 on Build GRCh38
rs762342196 Chr.6: 44254741-44254741 on Build GRCh38
rs751672960 Chr.6: 44254770-44254770 on Build GRCh38
rs758333497 Chr.6: 44254795-44254795 on Build GRCh38
rs766869254 Chr.6: 44254856-44254856 on Build GRCh38
rs540231426 Chr.6: 44254686-44254686 on Build GRCh38
rs756448430 Chr.6: 44254751-44254751 on Build GRCh38
rs140970584 Chr.6: 44254888-44254888 on Build GRCh38
rs202222313 Chr.6: 44254788-44254788 on Build GRCh38
rs781014560 Chr.6: 44254898-44254898 on Build GRCh38
rs375084737 Chr.6: 44254900-44254900 on Build GRCh38
rs200262124 Chr.6: 44254865-44254865 on Build GRCh38
rs764015490 Chr.6: 44254884-44254884 on Build GRCh38
rs372392256 Chr.6: 44254687-44254687 on Build GRCh38
rs755198190 Chr.6: 44254791-44254791 on Build GRCh38
rs757138296 Chr.6: 44254897-44254897 on Build GRCh38

View Full Product Details