Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143717984-143718249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK15
Entrez Gene ID
225689
Gene Name
mitogen-activated protein kinase 15
Gene Aliases
ERK7, ERK8
UniGene
Hs.493169
Gene Chromosome Location
Chr.8: 143716305-143722463 on Build GRCh38
Transcripts
NM_139021.2 XM_011516925.2 XM_011516926.2 XM_011516927.2 XM_011516928.2 XM_017013218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3924796 c.205G>A p.D69N Chr.8: 143718221-143718221 on Build GRCh38
5097691 c.231T>C p.L77L Chr.8: 143718247-143718247 on Build GRCh38
5097692 c.231T>C p.L77L Chr.8: 143718247-143718247 on Build GRCh38
5156906 c.191T>C p.L64P Chr.8: 143718072-143718072 on Build GRCh38
5156907 c.191T>C p.L64P Chr.8: 143718072-143718072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782755693 Chr.8: 143718182-143718182 on Build GRCh38
rs199893934 Chr.8: 143718054-143718054 on Build GRCh38
rs781971195 Chr.8: 143718124-143718124 on Build GRCh38
rs201192106 Chr.8: 143718044-143718044 on Build GRCh38
rs556281397 Chr.8: 143718156-143718156 on Build GRCh38
rs370367858 Chr.8: 143718039-143718039 on Build GRCh38
rs782537280 Chr.8: 143718096-143718096 on Build GRCh38
rs782229599 Chr.8: 143718222-143718222 on Build GRCh38
rs782181718 Chr.8: 143718169-143718169 on Build GRCh38
rs782589433 Chr.8: 143718022-143718022 on Build GRCh38
rs782221221 Chr.8: 143718005-143718005 on Build GRCh38
rs140665680 Chr.8: 143718068-143718068 on Build GRCh38
rs782328178 Chr.8: 143718171-143718171 on Build GRCh38
rs782005554 Chr.8: 143718052-143718052 on Build GRCh38
rs782248842 Chr.8: 143718061-143718062 on Build GRCh38
rs377080766 Chr.8: 143718202-143718202 on Build GRCh38
rs782390013 Chr.8: 143718021-143718021 on Build GRCh38
rs151117968 Chr.8: 143718047-143718047 on Build GRCh38
rs375599746 Chr.8: 143718026-143718026 on Build GRCh38
rs782072550 Chr.8: 143718062-143718062 on Build GRCh38
rs376917247 Chr.8: 143718191-143718191 on Build GRCh38
rs782168631 Chr.8: 143718172-143718172 on Build GRCh38
rs782266949 Chr.8: 143718105-143718105 on Build GRCh38
rs782751344 Chr.8: 143718193-143718193 on Build GRCh38
rs368058974 Chr.8: 143718198-143718198 on Build GRCh38
rs782347670 Chr.8: 143718111-143718111 on Build GRCh38
rs782476914 Chr.8: 143718075-143718075 on Build GRCh38
rs35285081 Chr.8: 143718066-143718066 on Build GRCh38
rs201928413 Chr.8: 143718004-143718004 on Build GRCh38
rs782520699 Chr.8: 143718214-143718214 on Build GRCh38
rs374120470 Chr.8: 143718094-143718094 on Build GRCh38
rs567179436 Chr.8: 143718187-143718187 on Build GRCh38
rs372524825 Chr.8: 143718012-143718012 on Build GRCh38
rs781815886 Chr.8: 143718073-143718073 on Build GRCh38
rs782634017 Chr.8: 143718102-143718102 on Build GRCh38
rs782090140 Chr.8: 143718192-143718192 on Build GRCh38
rs782576322 Chr.8: 143718083-143718083 on Build GRCh38
rs368587017 Chr.8: 143718221-143718221 on Build GRCh38
rs540839720 Chr.8: 143718137-143718137 on Build GRCh38
rs370910535 Chr.8: 143718197-143718197 on Build GRCh38
rs989637896 Chr.8: 143718123-143718123 on Build GRCh38
rs139756179 Chr.8: 143718057-143718057 on Build GRCh38
rs781854698 Chr.8: 143718205-143718205 on Build GRCh38
rs972166431 Chr.8: 143718055-143718055 on Build GRCh38
rs376145909 Chr.8: 143718129-143718129 on Build GRCh38
rs532189248 Chr.8: 143718130-143718130 on Build GRCh38
rs781795310 Chr.8: 143718061-143718061 on Build GRCh38

View Full Product Details