Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7484712-7484957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2A
Entrez Gene ID
5430
Gene Name
RNA polymerase II subunit A
Gene Aliases
POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
UniGene
Hs.270017
Gene Chromosome Location
Chr.17: 7484379-7514618 on Build GRCh38
Transcripts
NM_000937.4

SNP Detail

SNP ID Chromosome Location
rs750846594 Chr.17: 7484886-7484886 on Build GRCh38
rs1024560256 Chr.17: 7484792-7484792 on Build GRCh38
rs777199783 Chr.17: 7484778-7484779 on Build GRCh38
rs1001765651 Chr.17: 7484811-7484811 on Build GRCh38
rs765892388 Chr.17: 7484880-7484881 on Build GRCh38
rs942247212 Chr.17: 7484920-7484920 on Build GRCh38
rs772103266 Chr.17: 7484861-7484861 on Build GRCh38
rs369995545 Chr.17: 7484761-7484761 on Build GRCh38
rs190864669 Chr.17: 7484878-7484878 on Build GRCh38
rs756417344 Chr.17: 7484733-7484733 on Build GRCh38
rs768521150 Chr.17: 7484770-7484770 on Build GRCh38
rs757771437 Chr.17: 7484788-7484788 on Build GRCh38
rs769665843 Chr.17: 7484780-7484780 on Build GRCh38
rs976155358 Chr.17: 7484882-7484882 on Build GRCh38
rs757746668 Chr.17: 7484750-7484750 on Build GRCh38
rs751215469 Chr.17: 7484820-7484820 on Build GRCh38
rs1005262668 Chr.17: 7484777-7484777 on Build GRCh38
rs375984351 Chr.17: 7484932-7484932 on Build GRCh38
rs745428887 Chr.17: 7484758-7484758 on Build GRCh38
rs768926310 Chr.17: 7484876-7484876 on Build GRCh38
rs754306483 Chr.17: 7484747-7484747 on Build GRCh38
rs777501152 Chr.17: 7484840-7484840 on Build GRCh38
rs561641273 Chr.17: 7484763-7484763 on Build GRCh38
rs762376289 Chr.17: 7484779-7484779 on Build GRCh38
rs778465664 Chr.17: 7484754-7484754 on Build GRCh38
rs550849994 Chr.17: 7484901-7484901 on Build GRCh38
rs764468304 Chr.17: 7484746-7484746 on Build GRCh38
rs561861244 Chr.17: 7484885-7484885 on Build GRCh38
rs765732394 Chr.17: 7484798-7484798 on Build GRCh38
rs182661622 Chr.17: 7484897-7484897 on Build GRCh38
rs754205577 Chr.17: 7484787-7484787 on Build GRCh38
rs751162326 Chr.17: 7484906-7484906 on Build GRCh38
rs774472822 Chr.17: 7484781-7484781 on Build GRCh38
rs141361156 Chr.17: 7484827-7484827 on Build GRCh38
rs775645262 Chr.17: 7484785-7484785 on Build GRCh38
rs754661696 Chr.17: 7484907-7484907 on Build GRCh38
rs749274276 Chr.17: 7484860-7484860 on Build GRCh38
rs971220369 Chr.17: 7484802-7484802 on Build GRCh38
rs750944351 Chr.17: 7484800-7484800 on Build GRCh38
rs759598292 Chr.17: 7484782-7484782 on Build GRCh38
rs986702845 Chr.17: 7484903-7484903 on Build GRCh38
rs769761900 Chr.17: 7484833-7484833 on Build GRCh38
rs1036130754 Chr.17: 7484914-7484914 on Build GRCh38
rs553761464 Chr.17: 7484864-7484864 on Build GRCh38
rs967455777 Chr.17: 7484869-7484869 on Build GRCh38
rs764536293 Chr.17: 7484737-7484749 on Build GRCh38
rs893295621 Chr.17: 7484883-7484883 on Build GRCh38
rs973247410 Chr.17: 7484938-7484938 on Build GRCh38
rs867781905 Chr.17: 7484799-7484799 on Build GRCh38
rs746673121 Chr.17: 7484765-7484765 on Build GRCh38
rs373497066 Chr.17: 7484810-7484810 on Build GRCh38
rs112473664 Chr.17: 7484795-7484795 on Build GRCh38
rs776597121 Chr.17: 7484771-7484771 on Build GRCh38
rs138249788 Chr.17: 7484784-7484784 on Build GRCh38
rs913208975 Chr.17: 7484865-7484865 on Build GRCh38
rs760731040 Chr.17: 7484779-7484779 on Build GRCh38
rs757789292 Chr.17: 7484805-7484805 on Build GRCh38
rs762124073 Chr.17: 7484749-7484749 on Build GRCh38

View Full Product Details