Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44930431-44930668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PABPC1L
Entrez Gene ID
80336
Gene Name
poly(A) binding protein cytoplasmic 1 like
Gene Aliases
C20orf119, EPAB, PABPC1L1, dJ1069P2.3
UniGene
Hs.641481
Gene Chromosome Location
Chr.20: 44910060-44959160 on Build GRCh38
Transcripts
NM_001124756.2 NR_134983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7338215 c.1127C>T p.A376V Chr.20: 44930614-44930614 on Build GRCh38
7291809 c.1108C>T p.R370C Chr.20: 44930595-44930595 on Build GRCh38
7382181 c.1113A>T p.K371N Chr.20: 44930600-44930600 on Build GRCh38
6736647 c.1072G>A p.V358M Chr.20: 44930559-44930559 on Build GRCh38
7264713 c.1067G>A p.R356H Chr.20: 44930554-44930554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763497254 Chr.20: 44930573-44930573 on Build GRCh38
rs767814650 Chr.20: 44930641-44930641 on Build GRCh38
rs766822017 Chr.20: 44930579-44930579 on Build GRCh38
rs776479654 Chr.20: 44930626-44930626 on Build GRCh38
rs750480811 Chr.20: 44930567-44930567 on Build GRCh38
rs761027412 Chr.20: 44930452-44930452 on Build GRCh38
rs3746581 Chr.20: 44930558-44930558 on Build GRCh38
rs750947314 Chr.20: 44930524-44930524 on Build GRCh38
rs768627257 Chr.20: 44930621-44930621 on Build GRCh38
rs374768688 Chr.20: 44930634-44930634 on Build GRCh38
rs758897975 Chr.20: 44930525-44930525 on Build GRCh38
rs778336285 Chr.20: 44930607-44930607 on Build GRCh38
rs1048102523 Chr.20: 44930620-44930620 on Build GRCh38
rs894420771 Chr.20: 44930603-44930603 on Build GRCh38
rs746816385 Chr.20: 44930618-44930618 on Build GRCh38
rs765784961 Chr.20: 44930523-44930523 on Build GRCh38
rs553336356 Chr.20: 44930640-44930640 on Build GRCh38
rs762078937 Chr.20: 44930561-44930561 on Build GRCh38
rs369056714 Chr.20: 44930534-44930534 on Build GRCh38
rs764366575 Chr.20: 44930461-44930461 on Build GRCh38
rs372472103 Chr.20: 44930535-44930535 on Build GRCh38
rs774780363 Chr.20: 44930637-44930637 on Build GRCh38
rs778512925 Chr.20: 44930488-44930488 on Build GRCh38
rs751952360 Chr.20: 44930585-44930585 on Build GRCh38
rs780074379 Chr.20: 44930613-44930613 on Build GRCh38
rs770407127 Chr.20: 44930537-44930537 on Build GRCh38
rs763427943 Chr.20: 44930496-44930496 on Build GRCh38
rs748354804 Chr.20: 44930628-44930628 on Build GRCh38
rs776103554 Chr.20: 44930466-44930467 on Build GRCh38
rs778483862 Chr.20: 44930549-44930549 on Build GRCh38
rs756904215 Chr.20: 44930601-44930601 on Build GRCh38
rs201834135 Chr.20: 44930554-44930554 on Build GRCh38
rs755266874 Chr.20: 44930594-44930594 on Build GRCh38
rs765297788 Chr.20: 44930566-44930566 on Build GRCh38
rs907024701 Chr.20: 44930592-44930592 on Build GRCh38
rs924266395 Chr.20: 44930475-44930475 on Build GRCh38
rs373194491 Chr.20: 44930555-44930555 on Build GRCh38
rs370421458 Chr.20: 44930633-44930633 on Build GRCh38
rs773185715 Chr.20: 44930632-44930632 on Build GRCh38
rs368511144 Chr.20: 44930611-44930611 on Build GRCh38
rs376354337 Chr.20: 44930596-44930596 on Build GRCh38
rs577081130 Chr.20: 44930595-44930595 on Build GRCh38
rs764180149 Chr.20: 44930540-44930541 on Build GRCh38
rs770031609 Chr.20: 44930630-44930630 on Build GRCh38
rs753987524 Chr.20: 44930468-44930468 on Build GRCh38
rs199499305 Chr.20: 44930553-44930553 on Build GRCh38
rs745323605 Chr.20: 44930608-44930608 on Build GRCh38
rs753084519 Chr.20: 44930649-44930649 on Build GRCh38

View Full Product Details