Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 397578-397827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKP3
Entrez Gene ID
11187
Gene Name
plakophilin 3
Gene Aliases
-
UniGene
Hs.534395
Gene Chromosome Location
Chr.11: 392599-404908 on Build GRCh38
Transcripts
NM_001303029.1 NM_007183.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838515 c.1009G>A p.V337M Chr.11: 397603-397603 on Build GRCh38
7137621 c.1068G>A p.Q356Q Chr.11: 397662-397662 on Build GRCh38
1725502 c.1003C>G p.L335V Chr.11: 397597-397597 on Build GRCh38
3447192 c.1062G>A p.K354K Chr.11: 397656-397656 on Build GRCh38
4424975 c.1056C>T p.A352A Chr.11: 397650-397650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373109185 Chr.11: 397680-397680 on Build GRCh38
rs373906660 Chr.11: 397686-397686 on Build GRCh38
rs764364230 Chr.11: 397776-397776 on Build GRCh38
rs746797898 Chr.11: 397635-397635 on Build GRCh38
rs759112934 Chr.11: 397676-397676 on Build GRCh38
rs138662102 Chr.11: 397642-397642 on Build GRCh38
rs758587273 Chr.11: 397759-397759 on Build GRCh38
rs1043397755 Chr.11: 397774-397774 on Build GRCh38
rs148819261 Chr.11: 397651-397651 on Build GRCh38
rs774942363 Chr.11: 397650-397650 on Build GRCh38
rs749422728 Chr.11: 397693-397693 on Build GRCh38
rs777645595 Chr.11: 397623-397623 on Build GRCh38
rs911730416 Chr.11: 397725-397725 on Build GRCh38
rs3210555 Chr.11: 397609-397609 on Build GRCh38
rs757088043 Chr.11: 397692-397692 on Build GRCh38
rs149165268 Chr.11: 397760-397760 on Build GRCh38
rs146140233 Chr.11: 397614-397614 on Build GRCh38
rs753442379 Chr.11: 397604-397604 on Build GRCh38
rs1001730230 Chr.11: 397615-397615 on Build GRCh38
rs368726163 Chr.11: 397613-397613 on Build GRCh38
rs765938659 Chr.11: 397603-397603 on Build GRCh38
rs768577856 Chr.11: 397647-397647 on Build GRCh38
rs755096788 Chr.11: 397698-397698 on Build GRCh38
rs145544147 Chr.11: 397752-397752 on Build GRCh38
rs776362668 Chr.11: 397713-397713 on Build GRCh38
rs571337193 Chr.11: 397639-397639 on Build GRCh38
rs9795299 Chr.11: 397732-397732 on Build GRCh38
rs760116413 Chr.11: 397688-397688 on Build GRCh38
rs758285473 Chr.11: 397621-397621 on Build GRCh38
rs780117288 Chr.11: 397641-397641 on Build GRCh38
rs370191440 Chr.11: 397673-397673 on Build GRCh38
rs754418177 Chr.11: 397605-397605 on Build GRCh38
rs562426586 Chr.11: 397778-397778 on Build GRCh38
rs181543429 Chr.11: 397779-397779 on Build GRCh38
rs748219506 Chr.11: 397702-397702 on Build GRCh38
rs9795298 Chr.11: 397706-397706 on Build GRCh38
rs200512397 Chr.11: 397709-397709 on Build GRCh38
rs890965538 Chr.11: 397753-397753 on Build GRCh38
rs778790262 Chr.11: 397701-397701 on Build GRCh38
rs764115451 Chr.11: 397689-397689 on Build GRCh38
rs188555376 Chr.11: 397644-397644 on Build GRCh38
rs928824941 Chr.11: 397679-397679 on Build GRCh38
rs773683560 Chr.11: 397708-397708 on Build GRCh38
rs776110670 Chr.11: 397674-397674 on Build GRCh38
rs773630706 Chr.11: 397667-397667 on Build GRCh38
rs377205895 Chr.11: 397691-397691 on Build GRCh38
rs1052377908 Chr.11: 397741-397741 on Build GRCh38
rs149716601 Chr.11: 397685-397685 on Build GRCh38
rs368689289 Chr.11: 397750-397750 on Build GRCh38

View Full Product Details