Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45081422-45081693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACAD
Entrez Gene ID
23148
Gene Name
NAC alpha domain containing
Gene Aliases
-
UniGene
Hs.96633
Gene Chromosome Location
Chr.7: 45080432-45089443 on Build GRCh38
Transcripts
NM_001146334.1 XM_006715674.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1090039 c.4253G>A p.R1418Q Chr.7: 45081605-45081605 on Build GRCh38
3832876 c.4257+2T>G p.? Chr.7: 45081599-45081599 on Build GRCh38
3431624 c.4235G>A p.R1412H Chr.7: 45081623-45081623 on Build GRCh38
6212497 c.4257+4G>T p.? Chr.7: 45081597-45081597 on Build GRCh38
3263274 c.4197C>T p.G1399G Chr.7: 45081661-45081661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191131570 Chr.7: 45081648-45081648 on Build GRCh38
rs565592512 Chr.7: 45081457-45081457 on Build GRCh38
rs990831687 Chr.7: 45081463-45081463 on Build GRCh38
rs941679120 Chr.7: 45081639-45081644 on Build GRCh38
rs536193771 Chr.7: 45081485-45081485 on Build GRCh38
rs200858358 Chr.7: 45081642-45081642 on Build GRCh38
rs554335289 Chr.7: 45081506-45081506 on Build GRCh38
rs928479944 Chr.7: 45081530-45081530 on Build GRCh38
rs770548190 Chr.7: 45081636-45081636 on Build GRCh38
rs958066579 Chr.7: 45081455-45081455 on Build GRCh38
rs748965257 Chr.7: 45081598-45081598 on Build GRCh38
rs988986010 Chr.7: 45081531-45081531 on Build GRCh38
rs929234838 Chr.7: 45081534-45081534 on Build GRCh38
rs981312694 Chr.7: 45081507-45081507 on Build GRCh38
rs999775201 Chr.7: 45081664-45081664 on Build GRCh38
rs912869619 Chr.7: 45081560-45081560 on Build GRCh38
rs1032303421 Chr.7: 45081449-45081449 on Build GRCh38
rs997702842 Chr.7: 45081649-45081651 on Build GRCh38
rs1051840874 Chr.7: 45081643-45081643 on Build GRCh38
rs376697250 Chr.7: 45081654-45081654 on Build GRCh38
rs576219276 Chr.7: 45081638-45081638 on Build GRCh38
rs1048140788 Chr.7: 45081632-45081632 on Build GRCh38
rs776310654 Chr.7: 45081527-45081527 on Build GRCh38
rs796253381 Chr.7: 45081661-45081661 on Build GRCh38

View Full Product Details