Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16576157-16576416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED26
Entrez Gene ID
9441
Gene Name
mediator complex subunit 26
Gene Aliases
CRSP7, CRSP70
UniGene
Hs.715040
Gene Chromosome Location
Chr.19: 16574907-16628204 on Build GRCh38
Transcripts
NM_004831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7085736 c.1475G>A p.R492Q Chr.19: 16576355-16576355 on Build GRCh38
5781364 c.1594G>A p.V532M Chr.19: 16576236-16576236 on Build GRCh38
7124524 c.1537G>C p.E513Q Chr.19: 16576293-16576293 on Build GRCh38
5486008 c.1424C>T p.T475M Chr.19: 16576406-16576406 on Build GRCh38
3891804 c.1651G>A p.D551N Chr.19: 16576179-16576179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147206240 Chr.19: 16576266-16576266 on Build GRCh38
rs769468738 Chr.19: 16576311-16576311 on Build GRCh38
rs762876909 Chr.19: 16576331-16576331 on Build GRCh38
rs376850229 Chr.19: 16576265-16576265 on Build GRCh38
rs746642278 Chr.19: 16576381-16576381 on Build GRCh38
rs772758171 Chr.19: 16576221-16576221 on Build GRCh38
rs746368183 Chr.19: 16576259-16576259 on Build GRCh38
rs765030458 Chr.19: 16576176-16576176 on Build GRCh38
rs780507334 Chr.19: 16576355-16576355 on Build GRCh38
rs771751712 Chr.19: 16576287-16576287 on Build GRCh38
rs189485804 Chr.19: 16576196-16576196 on Build GRCh38
rs143751642 Chr.19: 16576197-16576197 on Build GRCh38
rs375832961 Chr.19: 16576216-16576216 on Build GRCh38
rs546631974 Chr.19: 16576339-16576339 on Build GRCh38
rs765871005 Chr.19: 16576223-16576223 on Build GRCh38
rs1000619860 Chr.19: 16576206-16576206 on Build GRCh38
rs781584589 Chr.19: 16576187-16576187 on Build GRCh38
rs139417299 Chr.19: 16576183-16576183 on Build GRCh38
rs777432485 Chr.19: 16576291-16576291 on Build GRCh38
rs1033751093 Chr.19: 16576271-16576271 on Build GRCh38
rs369798896 Chr.19: 16576326-16576326 on Build GRCh38
rs746622618 Chr.19: 16576315-16576315 on Build GRCh38
rs778270589 Chr.19: 16576273-16576275 on Build GRCh38
rs751746865 Chr.19: 16576336-16576336 on Build GRCh38
rs764943456 Chr.19: 16576232-16576232 on Build GRCh38
rs371790045 Chr.19: 16576242-16576242 on Build GRCh38
rs769837121 Chr.19: 16576330-16576330 on Build GRCh38
rs776453499 Chr.19: 16576228-16576228 on Build GRCh38
rs775766169 Chr.19: 16576215-16576215 on Build GRCh38
rs369433580 Chr.19: 16576386-16576386 on Build GRCh38
rs201082644 Chr.19: 16576179-16576179 on Build GRCh38
rs201627212 Chr.19: 16576244-16576244 on Build GRCh38
rs750503128 Chr.19: 16576345-16576345 on Build GRCh38
rs770610103 Chr.19: 16576385-16576385 on Build GRCh38
rs139574151 Chr.19: 16576327-16576327 on Build GRCh38
rs754302632 Chr.19: 16576356-16576356 on Build GRCh38
rs148231218 Chr.19: 16576177-16576177 on Build GRCh38
rs755356567 Chr.19: 16576363-16576363 on Build GRCh38
rs200454256 Chr.19: 16576222-16576222 on Build GRCh38
rs566314657 Chr.19: 16576341-16576341 on Build GRCh38
rs779188888 Chr.19: 16576366-16576366 on Build GRCh38
rs570538372 Chr.19: 16576295-16576295 on Build GRCh38
rs747067734 Chr.19: 16576180-16576180 on Build GRCh38
rs147359880 Chr.19: 16576182-16576182 on Build GRCh38
rs745357888 Chr.19: 16576392-16576392 on Build GRCh38
rs770358182 Chr.19: 16576324-16576324 on Build GRCh38
rs199739577 Chr.19: 16576325-16576325 on Build GRCh38
rs531804917 Chr.19: 16576243-16576243 on Build GRCh38
rs763935267 Chr.19: 16576333-16576333 on Build GRCh38
rs756629468 Chr.19: 16576352-16576352 on Build GRCh38
rs749630291 Chr.19: 16576269-16576269 on Build GRCh38
rs771700366 Chr.19: 16576217-16576217 on Build GRCh38
rs1045031545 Chr.19: 16576300-16576300 on Build GRCh38
rs752531834 Chr.19: 16576236-16576236 on Build GRCh38

View Full Product Details