Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108383514-108383787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH15
Entrez Gene ID
22989
Gene Name
myosin heavy chain 15
Gene Aliases
-
UniGene
Hs.731060
Gene Chromosome Location
Chr.3: 108380369-108549462 on Build GRCh38
Transcripts
NM_014981.1 XM_011512559.2 XM_017005922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5431444 c.5692-4A>T p.? Chr.3: 108383733-108383733 on Build GRCh38
6468280 c.5749G>A p.E1917K Chr.3: 108383672-108383672 on Build GRCh38
3914472 c.5692G>A p.E1898K Chr.3: 108383729-108383729 on Build GRCh38
6252420 c.5765C>A p.A1922E Chr.3: 108383656-108383656 on Build GRCh38
5934790 c.5692-1G>A p.? Chr.3: 108383730-108383730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372472456 Chr.3: 108383746-108383747 on Build GRCh38
rs776793750 Chr.3: 108383724-108383724 on Build GRCh38
rs775690523 Chr.3: 108383755-108383755 on Build GRCh38
rs750670497 Chr.3: 108383742-108383743 on Build GRCh38
rs138733417 Chr.3: 108383738-108383739 on Build GRCh38
rs766203154 Chr.3: 108383741-108383741 on Build GRCh38
rs763291537 Chr.3: 108383730-108383730 on Build GRCh38
rs747054348 Chr.3: 108383722-108383722 on Build GRCh38
rs953600514 Chr.3: 108383548-108383548 on Build GRCh38
rs758864849 Chr.3: 108383722-108383723 on Build GRCh38
rs746003362 Chr.3: 108383747-108383747 on Build GRCh38
rs750532615 Chr.3: 108383735-108383736 on Build GRCh38
rs780461864 Chr.3: 108383725-108383726 on Build GRCh38
rs796146085 Chr.3: 108383738-108383739 on Build GRCh38
rs377049870 Chr.3: 108383733-108383734 on Build GRCh38
rs200773679 Chr.3: 108383600-108383600 on Build GRCh38
rs747835756 Chr.3: 108383732-108383737 on Build GRCh38
rs753622607 Chr.3: 108383742-108383742 on Build GRCh38
rs755817806 Chr.3: 108383740-108383740 on Build GRCh38
rs547582039 Chr.3: 108383756-108383756 on Build GRCh38
rs767963441 Chr.3: 108383745-108383746 on Build GRCh38
rs397818948 Chr.3: 108383754-108383755 on Build GRCh38
rs79837783 Chr.3: 108383733-108383733 on Build GRCh38
rs756722476 Chr.3: 108383654-108383654 on Build GRCh38
rs750135705 Chr.3: 108383664-108383664 on Build GRCh38
rs758399754 Chr.3: 108383744-108383744 on Build GRCh38
rs373566058 Chr.3: 108383748-108383748 on Build GRCh38
rs369307732 Chr.3: 108383687-108383687 on Build GRCh38
rs772062112 Chr.3: 108383750-108383751 on Build GRCh38
rs780923198 Chr.3: 108383731-108383732 on Build GRCh38
rs768636537 Chr.3: 108383720-108383720 on Build GRCh38
rs141454667 Chr.3: 108383737-108383738 on Build GRCh38
rs755594117 Chr.3: 108383744-108383745 on Build GRCh38
rs758237765 Chr.3: 108383610-108383610 on Build GRCh38
rs78006088 Chr.3: 108383723-108383723 on Build GRCh38
rs781225468 Chr.3: 108383746-108383746 on Build GRCh38
rs780834351 Chr.3: 108383660-108383660 on Build GRCh38
rs7426463 Chr.3: 108383739-108383739 on Build GRCh38
rs759844449 Chr.3: 108383603-108383603 on Build GRCh38
rs1034102406 Chr.3: 108383566-108383566 on Build GRCh38
rs745769427 Chr.3: 108383749-108383750 on Build GRCh38
rs775248561 Chr.3: 108383751-108383752 on Build GRCh38
rs559813965 Chr.3: 108383652-108383652 on Build GRCh38
rs972100592 Chr.3: 108383688-108383688 on Build GRCh38
rs749155812 Chr.3: 108383719-108383719 on Build GRCh38
rs770049114 Chr.3: 108383729-108383729 on Build GRCh38
rs1024388366 Chr.3: 108383582-108383582 on Build GRCh38
rs1013962640 Chr.3: 108383578-108383578 on Build GRCh38
rs199682590 Chr.3: 108383632-108383632 on Build GRCh38
rs368208731 Chr.3: 108383739-108383739 on Build GRCh38
rs746747679 Chr.3: 108383752-108383755 on Build GRCh38
rs747390211 Chr.3: 108383729-108383729 on Build GRCh38
rs200099329 Chr.3: 108383736-108383737 on Build GRCh38
rs766510245 Chr.3: 108383743-108383744 on Build GRCh38
rs765620642 Chr.3: 108383614-108383614 on Build GRCh38
rs761334625 Chr.3: 108383732-108383732 on Build GRCh38
rs770844553 Chr.3: 108383733-108383733 on Build GRCh38
rs765500062 Chr.3: 108383741-108383742 on Build GRCh38
rs779976121 Chr.3: 108383714-108383714 on Build GRCh38
rs768526867 Chr.3: 108383570-108383570 on Build GRCh38
rs750915570 Chr.3: 108383594-108383594 on Build GRCh38
rs778861932 Chr.3: 108383743-108383743 on Build GRCh38
rs778530962 Chr.3: 108383748-108383749 on Build GRCh38
rs764564051 Chr.3: 108383636-108383636 on Build GRCh38
rs760734119 Chr.3: 108383576-108383576 on Build GRCh38
rs762068316 Chr.3: 108383735-108383739 on Build GRCh38
rs80293068 Chr.3: 108383594-108383594 on Build GRCh38
rs773331383 Chr.3: 108383573-108383573 on Build GRCh38
rs756686160 Chr.3: 108383747-108383748 on Build GRCh38
rs1016823703 Chr.3: 108383661-108383661 on Build GRCh38
rs746058501 Chr.3: 108383725-108383725 on Build GRCh38
rs768499828 Chr.3: 108383752-108383753 on Build GRCh38
rs769154472 Chr.3: 108383730-108383731 on Build GRCh38
rs759536734 Chr.3: 108383740-108383740 on Build GRCh38
rs951353754 Chr.3: 108383616-108383621 on Build GRCh38
rs774730236 Chr.3: 108383731-108383731 on Build GRCh38
rs753099471 Chr.3: 108383628-108383628 on Build GRCh38
rs752109758 Chr.3: 108383737-108383739 on Build GRCh38
rs764981322 Chr.3: 108383739-108383740 on Build GRCh38
rs758924170 Chr.3: 108383736-108383739 on Build GRCh38
rs34041576 Chr.3: 108383740-108383741 on Build GRCh38
rs765266302 Chr.3: 108383734-108383735 on Build GRCh38

View Full Product Details