Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121033512-121033739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OASL
Entrez Gene ID
8638
Gene Name
2'-5'-oligoadenylate synthetase like
Gene Aliases
OASLd, TRIP-14, TRIP14, p59 OASL, p59-OASL, p59OASL
UniGene
Hs.118633
Gene Chromosome Location
Chr.12: 121018882-121039242 on Build GRCh38
Transcripts
NM_001261825.1 NM_003733.3 NM_198213.2 XM_017020140.1 XM_017020141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700223 c.312T>C p.V104V Chr.12: 121033630-121033630 on Build GRCh38
3670992 c.211G>T p.G71W Chr.12: 121033731-121033731 on Build GRCh38
691754 c.242A>C p.E81A Chr.12: 121033700-121033700 on Build GRCh38
3457128 c.239G>A p.R80K Chr.12: 121033703-121033703 on Build GRCh38
1359424 c.331delA p.T111fs*16 Chr.12: 121033611-121033611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765440561 Chr.12: 121033684-121033684 on Build GRCh38
rs774039173 Chr.12: 121033671-121033673 on Build GRCh38
rs766041481 Chr.12: 121033553-121033554 on Build GRCh38
rs760031887 Chr.12: 121033543-121033543 on Build GRCh38
rs761944435 Chr.12: 121033668-121033668 on Build GRCh38
rs754861811 Chr.12: 121033591-121033591 on Build GRCh38
rs892136617 Chr.12: 121033580-121033588 on Build GRCh38
rs755737507 Chr.12: 121033722-121033722 on Build GRCh38
rs373574251 Chr.12: 121033720-121033720 on Build GRCh38
rs762437111 Chr.12: 121033548-121033549 on Build GRCh38
rs140204273 Chr.12: 121033538-121033538 on Build GRCh38
rs776013396 Chr.12: 121033654-121033654 on Build GRCh38
rs568732289 Chr.12: 121033608-121033608 on Build GRCh38
rs763085279 Chr.12: 121033695-121033695 on Build GRCh38
rs1000567798 Chr.12: 121033627-121033627 on Build GRCh38
rs777156196 Chr.12: 121033622-121033622 on Build GRCh38
rs145474725 Chr.12: 121033579-121033579 on Build GRCh38
rs772766111 Chr.12: 121033541-121033541 on Build GRCh38
rs1032407484 Chr.12: 121033649-121033649 on Build GRCh38
rs752473513 Chr.12: 121033721-121033721 on Build GRCh38
rs553252644 Chr.12: 121033563-121033563 on Build GRCh38
rs143025848 Chr.12: 121033561-121033561 on Build GRCh38
rs767950583 Chr.12: 121033550-121033550 on Build GRCh38
rs765299664 Chr.12: 121033572-121033572 on Build GRCh38
rs368869842 Chr.12: 121033709-121033709 on Build GRCh38
rs750491713 Chr.12: 121033692-121033692 on Build GRCh38
rs768970379 Chr.12: 121033617-121033617 on Build GRCh38
rs769486678 Chr.12: 121033609-121033609 on Build GRCh38
rs776384404 Chr.12: 121033535-121033535 on Build GRCh38
rs746832678 Chr.12: 121033586-121033586 on Build GRCh38
rs777046945 Chr.12: 121033666-121033666 on Build GRCh38
rs780781838 Chr.12: 121033600-121033600 on Build GRCh38
rs148845781 Chr.12: 121033656-121033656 on Build GRCh38
rs754052081 Chr.12: 121033564-121033564 on Build GRCh38
rs772594511 Chr.12: 121033646-121033646 on Build GRCh38
rs149658390 Chr.12: 121033560-121033560 on Build GRCh38
rs866108869 Chr.12: 121033685-121033685 on Build GRCh38
rs766537169 Chr.12: 121033699-121033699 on Build GRCh38
rs3213545 Chr.12: 121033534-121033534 on Build GRCh38
rs769065053 Chr.12: 121033665-121033665 on Build GRCh38
rs751596317 Chr.12: 121033707-121033707 on Build GRCh38
rs374804957 Chr.12: 121033585-121033585 on Build GRCh38
rs757485079 Chr.12: 121033565-121033565 on Build GRCh38
rs748742761 Chr.12: 121033641-121033641 on Build GRCh38

View Full Product Details