Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75012331-75012568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARM1
Entrez Gene ID
25849
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
Cipar1, DKFZP564O0823, PARM-1, WSC4
UniGene
Hs.105460
Gene Chromosome Location
Chr.4: 74933075-75050115 on Build GRCh38
Transcripts
NM_015393.3 XM_011531833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1430994 c.155A>G p.D52G Chr.4: 75012536-75012536 on Build GRCh38
3775980 c.126G>T p.W42C Chr.4: 75012507-75012507 on Build GRCh38
1430988 c.96G>A p.P32P Chr.4: 75012477-75012477 on Build GRCh38
5043521 c.351C>A p.N117K Chr.4: 75012555-75012555 on Build GRCh38
1430985 c.95C>T p.P32L Chr.4: 75012476-75012476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761427988 Chr.4: 75012426-75012426 on Build GRCh38
rs578038379 Chr.4: 75012498-75012498 on Build GRCh38
rs749936199 Chr.4: 75012453-75012453 on Build GRCh38
rs200338345 Chr.4: 75012527-75012527 on Build GRCh38
rs754987524 Chr.4: 75012477-75012477 on Build GRCh38
rs1011920106 Chr.4: 75012549-75012549 on Build GRCh38
rs776739917 Chr.4: 75012389-75012389 on Build GRCh38
rs963338676 Chr.4: 75012548-75012548 on Build GRCh38
rs760390554 Chr.4: 75012543-75012543 on Build GRCh38
rs929419093 Chr.4: 75012539-75012539 on Build GRCh38
rs774680116 Chr.4: 75012530-75012530 on Build GRCh38
rs769194001 Chr.4: 75012529-75012529 on Build GRCh38
rs751954500 Chr.4: 75012418-75012418 on Build GRCh38
rs775926286 Chr.4: 75012407-75012407 on Build GRCh38
rs199499699 Chr.4: 75012531-75012531 on Build GRCh38
rs763413479 Chr.4: 75012528-75012528 on Build GRCh38
rs759580161 Chr.4: 75012405-75012405 on Build GRCh38
rs915896469 Chr.4: 75012525-75012525 on Build GRCh38
rs781323245 Chr.4: 75012495-75012495 on Build GRCh38
rs763452456 Chr.4: 75012413-75012413 on Build GRCh38
rs938774329 Chr.4: 75012491-75012491 on Build GRCh38
rs770317662 Chr.4: 75012406-75012406 on Build GRCh38
rs923831009 Chr.4: 75012374-75012374 on Build GRCh38
rs769694183 Chr.4: 75012520-75012520 on Build GRCh38
rs533961402 Chr.4: 75012494-75012494 on Build GRCh38
rs149171425 Chr.4: 75012381-75012381 on Build GRCh38
rs958231139 Chr.4: 75012414-75012414 on Build GRCh38
rs993260611 Chr.4: 75012365-75012365 on Build GRCh38
rs1030061319 Chr.4: 75012367-75012367 on Build GRCh38
rs747179918 Chr.4: 75012382-75012382 on Build GRCh38
rs942289823 Chr.4: 75012526-75012526 on Build GRCh38
rs771307587 Chr.4: 75012383-75012383 on Build GRCh38
rs749977694 Chr.4: 75012536-75012536 on Build GRCh38
rs755647341 Chr.4: 75012461-75012461 on Build GRCh38
rs371114006 Chr.4: 75012479-75012479 on Build GRCh38
rs377545871 Chr.4: 75012466-75012466 on Build GRCh38
rs772034017 Chr.4: 75012520-75012520 on Build GRCh38
rs373919103 Chr.4: 75012441-75012441 on Build GRCh38
rs182618057 Chr.4: 75012476-75012476 on Build GRCh38
rs767195106 Chr.4: 75012533-75012533 on Build GRCh38

View Full Product Details