Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144466970-144467211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIFC2
Entrez Gene ID
90990
Gene Name
kinesin family member C2
Gene Aliases
-
UniGene
Hs.528713
Gene Chromosome Location
Chr.8: 144466246-144474116 on Build GRCh38
Transcripts
NM_145754.3 XM_005272357.3 XM_011517362.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6695763 c.339G>A p.E113E Chr.8: 144467211-144467211 on Build GRCh38
6180500 c.232G>A p.A78T Chr.8: 144467012-144467012 on Build GRCh38
4808133 c.279C>T p.S93S Chr.8: 144467059-144467059 on Build GRCh38
3750122 c.222C>T p.G74G Chr.8: 144467002-144467002 on Build GRCh38
2871705 c.282C>G p.V94V Chr.8: 144467062-144467062 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759548872 Chr.8: 144467041-144467041 on Build GRCh38
rs775638519 Chr.8: 144467109-144467109 on Build GRCh38
rs780492551 Chr.8: 144467062-144467062 on Build GRCh38
rs966494425 Chr.8: 144466988-144466988 on Build GRCh38
rs202237016 Chr.8: 144467194-144467194 on Build GRCh38
rs553351396 Chr.8: 144467094-144467094 on Build GRCh38
rs2250891 Chr.8: 144467002-144467002 on Build GRCh38
rs776381795 Chr.8: 144467039-144467039 on Build GRCh38
rs746540082 Chr.8: 144467133-144467133 on Build GRCh38
rs898567839 Chr.8: 144467092-144467092 on Build GRCh38
rs747649041 Chr.8: 144467074-144467074 on Build GRCh38
rs201988058 Chr.8: 144467137-144467137 on Build GRCh38
rs769483233 Chr.8: 144467141-144467141 on Build GRCh38
rs767849226 Chr.8: 144467058-144467058 on Build GRCh38
rs377030636 Chr.8: 144467059-144467059 on Build GRCh38
rs372835242 Chr.8: 144467187-144467187 on Build GRCh38
rs756784364 Chr.8: 144467157-144467157 on Build GRCh38
rs201345666 Chr.8: 144467152-144467152 on Build GRCh38
rs777346077 Chr.8: 144467079-144467079 on Build GRCh38
rs895098195 Chr.8: 144467130-144467130 on Build GRCh38
rs770275365 Chr.8: 144467036-144467036 on Build GRCh38
rs754213178 Chr.8: 144467122-144467122 on Build GRCh38
rs779450037 Chr.8: 144467073-144467073 on Build GRCh38
rs762058316 Chr.8: 144466999-144467013 on Build GRCh38
rs200941622 Chr.8: 144467181-144467181 on Build GRCh38
rs778520731 Chr.8: 144467027-144467027 on Build GRCh38
rs759706809 Chr.8: 144467188-144467188 on Build GRCh38
rs200556009 Chr.8: 144467166-144467166 on Build GRCh38
rs777294710 Chr.8: 144467132-144467132 on Build GRCh38
rs781614098 Chr.8: 144467011-144467011 on Build GRCh38
rs370533406 Chr.8: 144467177-144467177 on Build GRCh38
rs760993287 Chr.8: 144467118-144467118 on Build GRCh38
rs780361524 Chr.8: 144467020-144467020 on Build GRCh38
rs764256709 Chr.8: 144467003-144467003 on Build GRCh38
rs757523173 Chr.8: 144467007-144467007 on Build GRCh38
rs747039996 Chr.8: 144467179-144467179 on Build GRCh38
rs913683527 Chr.8: 144467010-144467010 on Build GRCh38
rs770042258 Chr.8: 144467191-144467191 on Build GRCh38
rs766552844 Chr.8: 144467121-144467121 on Build GRCh38
rs957680349 Chr.8: 144467103-144467103 on Build GRCh38
rs768007332 Chr.8: 144467023-144467023 on Build GRCh38
rs753815473 Chr.8: 144466990-144466990 on Build GRCh38
rs746684667 Chr.8: 144467087-144467087 on Build GRCh38
rs760392294 Chr.8: 144467035-144467035 on Build GRCh38
rs771696200 Chr.8: 144467033-144467033 on Build GRCh38
rs750806320 Chr.8: 144467019-144467019 on Build GRCh38
rs771571926 Chr.8: 144467078-144467078 on Build GRCh38
rs751067506 Chr.8: 144467156-144467156 on Build GRCh38
rs773399867 Chr.8: 144467147-144467147 on Build GRCh38
rs755482942 Chr.8: 144467066-144467066 on Build GRCh38
rs760720082 Chr.8: 144467149-144467149 on Build GRCh38
rs372510527 Chr.8: 144467175-144467175 on Build GRCh38
rs749817940 Chr.8: 144467021-144467021 on Build GRCh38
rs200097391 Chr.8: 144467090-144467090 on Build GRCh38
rs773606447 Chr.8: 144467115-144467115 on Build GRCh38
rs368346790 Chr.8: 144467151-144467151 on Build GRCh38
rs557479300 Chr.8: 144467135-144467135 on Build GRCh38
rs760666284 Chr.8: 144467171-144467171 on Build GRCh38
rs764634020 Chr.8: 144467154-144467154 on Build GRCh38
rs199946114 Chr.8: 144467138-144467138 on Build GRCh38
rs535998933 Chr.8: 144467127-144467127 on Build GRCh38
rs753212230 Chr.8: 144467128-144467128 on Build GRCh38
rs755926640 Chr.8: 144467169-144467169 on Build GRCh38
rs772241513 Chr.8: 144467144-144467144 on Build GRCh38
rs539596910 Chr.8: 144467143-144467143 on Build GRCh38
rs751667885 Chr.8: 144467053-144467053 on Build GRCh38
rs765126060 Chr.8: 144467047-144467047 on Build GRCh38
rs373065293 Chr.8: 144467006-144467006 on Build GRCh38
rs571105762 Chr.8: 144467088-144467088 on Build GRCh38
rs754264411 Chr.8: 144467063-144467063 on Build GRCh38
rs772349171 Chr.8: 144467010-144467034 on Build GRCh38
rs1032293883 Chr.8: 144467098-144467098 on Build GRCh38
rs370445562 Chr.8: 144467125-144467125 on Build GRCh38
rs757853816 Chr.8: 144467129-144467129 on Build GRCh38
rs766616444 Chr.8: 144467150-144467150 on Build GRCh38
rs540115052 Chr.8: 144467192-144467192 on Build GRCh38
rs1032529202 Chr.8: 144467145-144467145 on Build GRCh38
rs375661122 Chr.8: 144467176-144467176 on Build GRCh38
rs756378315 Chr.8: 144467060-144467060 on Build GRCh38
rs749139339 Chr.8: 144467174-144467174 on Build GRCh38
rs905027580 Chr.8: 144467093-144467093 on Build GRCh38
rs772912756 Chr.8: 144467034-144467034 on Build GRCh38
rs1025194769 Chr.8: 144467065-144467065 on Build GRCh38
rs773679048 Chr.8: 144467124-144467125 on Build GRCh38
rs369226997 Chr.8: 144467165-144467165 on Build GRCh38
rs773520850 Chr.8: 144467117-144467117 on Build GRCh38
rs946217596 Chr.8: 144467015-144467015 on Build GRCh38
rs112027207 Chr.8: 144467014-144467014 on Build GRCh38
rs758869461 Chr.8: 144467153-144467153 on Build GRCh38
rs565074636 Chr.8: 144467032-144467032 on Build GRCh38
rs1037497240 Chr.8: 144467086-144467086 on Build GRCh38

View Full Product Details