Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111015663-111015920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPF2
Entrez Gene ID
84154
Gene Name
ribosome production factor 2 homolog
Gene Aliases
BXDC1, bA397G5.4
UniGene
Hs.372265
Gene Chromosome Location
Chr.6: 110982017-111028263 on Build GRCh38
Transcripts
NM_001289111.1 NM_032194.2 XM_017011354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857526 c.409C>T p.R137C Chr.6: 111015786-111015786 on Build GRCh38
3857524 c.408C>T p.I136I Chr.6: 111015785-111015785 on Build GRCh38
4421140 c.464A>G p.Y155C Chr.6: 111015841-111015841 on Build GRCh38
4632387 c.528C>T p.R176R Chr.6: 111015788-111015788 on Build GRCh38
5404164 c.469C>T p.R157* Chr.6: 111015846-111015846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932622054 Chr.6: 111015847-111015847 on Build GRCh38
rs749211309 Chr.6: 111015739-111015739 on Build GRCh38
rs1002810054 Chr.6: 111015735-111015735 on Build GRCh38
rs150251680 Chr.6: 111015787-111015787 on Build GRCh38
rs777571365 Chr.6: 111015732-111015732 on Build GRCh38
rs953220212 Chr.6: 111015891-111015891 on Build GRCh38
rs777468605 Chr.6: 111015863-111015863 on Build GRCh38
rs754486291 Chr.6: 111015814-111015814 on Build GRCh38
rs4947093 Chr.6: 111015875-111015875 on Build GRCh38
rs762039879 Chr.6: 111015780-111015780 on Build GRCh38
rs770396481 Chr.6: 111015704-111015704 on Build GRCh38
rs866358250 Chr.6: 111015813-111015813 on Build GRCh38
rs766952895 Chr.6: 111015831-111015831 on Build GRCh38
rs374167220 Chr.6: 111015708-111015708 on Build GRCh38
rs759412599 Chr.6: 111015712-111015713 on Build GRCh38
rs752436503 Chr.6: 111015857-111015858 on Build GRCh38
rs983859943 Chr.6: 111015743-111015743 on Build GRCh38
rs751130342 Chr.6: 111015806-111015806 on Build GRCh38
rs1019920785 Chr.6: 111015808-111015808 on Build GRCh38
rs149831797 Chr.6: 111015715-111015715 on Build GRCh38
rs202181813 Chr.6: 111015786-111015786 on Build GRCh38
rs374083143 Chr.6: 111015872-111015872 on Build GRCh38
rs761969490 Chr.6: 111015700-111015700 on Build GRCh38
rs967052217 Chr.6: 111015825-111015825 on Build GRCh38
rs766852387 Chr.6: 111015792-111015792 on Build GRCh38
rs751042387 Chr.6: 111015706-111015706 on Build GRCh38
rs780812281 Chr.6: 111015712-111015712 on Build GRCh38
rs145863310 Chr.6: 111015836-111015836 on Build GRCh38
rs760822408 Chr.6: 111015773-111015773 on Build GRCh38
rs769487414 Chr.6: 111015724-111015724 on Build GRCh38
rs770869665 Chr.6: 111015746-111015746 on Build GRCh38
rs755748875 Chr.6: 111015861-111015861 on Build GRCh38
rs765543807 Chr.6: 111015784-111015784 on Build GRCh38
rs769064933 Chr.6: 111015776-111015776 on Build GRCh38
rs143958546 Chr.6: 111015777-111015777 on Build GRCh38
rs775761405 Chr.6: 111015770-111015770 on Build GRCh38
rs776678486 Chr.6: 111015708-111015708 on Build GRCh38
rs765179384 Chr.6: 111015715-111015719 on Build GRCh38

View Full Product Details