Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31141512-31141767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS36
Entrez Gene ID
146547
Gene Name
protease, serine 36
Gene Aliases
-
UniGene
Hs.256632
Gene Chromosome Location
Chr.16: 31138926-31150094 on Build GRCh38
Transcripts
NM_001258290.1 NM_001258291.1 NM_173502.4 XM_011545758.2 XM_017022968.1 XM_017022969.1 XM_017022970.1 XM_017022971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1377609 c.1723G>T p.E575* Chr.16: 31141759-31141759 on Build GRCh38
5961196 c.1716G>C p.E572D Chr.16: 31141766-31141766 on Build GRCh38
6223636 c.1779G>A p.E593E Chr.16: 31141591-31141591 on Build GRCh38
4773218 c.1840G>T p.V614F Chr.16: 31141530-31141530 on Build GRCh38
1377607 c.1837C>T p.R613* Chr.16: 31141533-31141533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749978791 Chr.16: 31141719-31141719 on Build GRCh38
rs752568776 Chr.16: 31141547-31141547 on Build GRCh38
rs745514179 Chr.16: 31141721-31141722 on Build GRCh38
rs527934076 Chr.16: 31141675-31141675 on Build GRCh38
rs770144613 Chr.16: 31141688-31141689 on Build GRCh38
rs773231251 Chr.16: 31141530-31141530 on Build GRCh38
rs753602448 Chr.16: 31141713-31141713 on Build GRCh38
rs189621061 Chr.16: 31141727-31141727 on Build GRCh38
rs763073739 Chr.16: 31141533-31141533 on Build GRCh38
rs775399704 Chr.16: 31141694-31141694 on Build GRCh38
rs767160836 Chr.16: 31141538-31141538 on Build GRCh38
rs759326833 Chr.16: 31141692-31141692 on Build GRCh38
rs760583158 Chr.16: 31141700-31141700 on Build GRCh38
rs755512328 Chr.16: 31141610-31141610 on Build GRCh38
rs567100727 Chr.16: 31141668-31141668 on Build GRCh38
rs367979107 Chr.16: 31141680-31141680 on Build GRCh38
rs894261691 Chr.16: 31141729-31141729 on Build GRCh38
rs763829301 Chr.16: 31141712-31141712 on Build GRCh38
rs951361732 Chr.16: 31141532-31141532 on Build GRCh38
rs138969109 Chr.16: 31141626-31141626 on Build GRCh38
rs150228670 Chr.16: 31141535-31141535 on Build GRCh38
rs924887142 Chr.16: 31141740-31141740 on Build GRCh38
rs756946953 Chr.16: 31141716-31141716 on Build GRCh38
rs769982905 Chr.16: 31141647-31141647 on Build GRCh38
rs897815907 Chr.16: 31141581-31141581 on Build GRCh38
rs145749002 Chr.16: 31141599-31141599 on Build GRCh38
rs8053474 Chr.16: 31141632-31141632 on Build GRCh38
rs976129957 Chr.16: 31141598-31141598 on Build GRCh38
rs755776611 Chr.16: 31141549-31141549 on Build GRCh38
rs201184372 Chr.16: 31141687-31141687 on Build GRCh38
rs773463105 Chr.16: 31141648-31141648 on Build GRCh38
rs755392167 Chr.16: 31141726-31141726 on Build GRCh38
rs763884390 Chr.16: 31141571-31141571 on Build GRCh38
rs145938370 Chr.16: 31141552-31141552 on Build GRCh38
rs139120097 Chr.16: 31141536-31141536 on Build GRCh38
rs756538144 Chr.16: 31141732-31141732 on Build GRCh38
rs748481334 Chr.16: 31141731-31141731 on Build GRCh38
rs777963110 Chr.16: 31141738-31141738 on Build GRCh38
rs756894288 Chr.16: 31141587-31141587 on Build GRCh38
rs780596988 Chr.16: 31141590-31141590 on Build GRCh38
rs762192303 Chr.16: 31141628-31141628 on Build GRCh38
rs762966250 Chr.16: 31141745-31141745 on Build GRCh38
rs753363315 Chr.16: 31141582-31141582 on Build GRCh38
rs143680005 Chr.16: 31141537-31141537 on Build GRCh38
rs764582498 Chr.16: 31141717-31141717 on Build GRCh38
rs976314766 Chr.16: 31141698-31141698 on Build GRCh38

View Full Product Details