Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13735263-13735524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OFD1
Entrez Gene ID
8481
Gene Name
OFD1, centriole and centriolar satellite protein
Gene Aliases
71-7A, CXorf5, JBTS10, RP23, SGBS2
UniGene
Hs.6483
Gene Chromosome Location
Chr.X: 13734713-13769361 on Build GRCh38
Transcripts
NM_003611.2 XM_005274604.2 XM_005274606.3 XM_011545592.2 XM_011545594.2 XM_011545595.1 XM_011545597.1 XM_017029909.1 XM_017029910.1

SNP Detail

SNP ID Chromosome Location
rs189544035 Chr.X: 13735451-13735451 on Build GRCh38
rs1050332905 Chr.X: 13735336-13735336 on Build GRCh38
rs925091188 Chr.X: 13735354-13735354 on Build GRCh38
rs777879661 Chr.X: 13735322-13735322 on Build GRCh38
rs964417549 Chr.X: 13735406-13735406 on Build GRCh38
rs886039857 Chr.X: 13735287-13735287 on Build GRCh38
rs184392923 Chr.X: 13735385-13735385 on Build GRCh38
rs312262807 Chr.X: 13735300-13735301 on Build GRCh38
rs975570918 Chr.X: 13735408-13735408 on Build GRCh38
rs374672797 Chr.X: 13735391-13735391 on Build GRCh38
rs312262809 Chr.X: 13735348-13735348 on Build GRCh38
rs886041737 Chr.X: 13735307-13735307 on Build GRCh38
rs940025902 Chr.X: 13735481-13735481 on Build GRCh38
rs908790070 Chr.X: 13735477-13735477 on Build GRCh38
rs756406468 Chr.X: 13735319-13735319 on Build GRCh38
rs921393501 Chr.X: 13735414-13735414 on Build GRCh38
rs1013221062 Chr.X: 13735448-13735448 on Build GRCh38
rs1018673918 Chr.X: 13735395-13735395 on Build GRCh38
rs749480884 Chr.X: 13735341-13735341 on Build GRCh38
rs312262804 Chr.X: 13735349-13735349 on Build GRCh38
rs371501200 Chr.X: 13735368-13735368 on Build GRCh38
rs312262808 Chr.X: 13735346-13735346 on Build GRCh38
rs1046113737 Chr.X: 13735484-13735484 on Build GRCh38
rs886039856 Chr.X: 13735297-13735298 on Build GRCh38
rs987095890 Chr.X: 13735390-13735390 on Build GRCh38
rs984812684 Chr.X: 13735455-13735455 on Build GRCh38
rs367927336 Chr.X: 13735329-13735329 on Build GRCh38
rs147114577 Chr.X: 13735289-13735289 on Build GRCh38
rs931590270 Chr.X: 13735447-13735447 on Build GRCh38
rs772705964 Chr.X: 13735392-13735392 on Build GRCh38

View Full Product Details