Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241548640-241548903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KMO
Entrez Gene ID
8564
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Gene Aliases
dJ317G22.1
UniGene
Hs.731056
Gene Chromosome Location
Chr.1: 241531883-241595647 on Build GRCh38
Transcripts
NM_003679.4 XM_005273337.1 XM_005273338.3 XM_017002649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417047 c.60C>T p.G20G Chr.1: 241548834-241548834 on Build GRCh38
5705350 c.124+1G>C p.? Chr.1: 241548899-241548899 on Build GRCh38
679713 c.90G>C p.R30S Chr.1: 241548864-241548864 on Build GRCh38
1626933 c.55-9A>G p.? Chr.1: 241548820-241548820 on Build GRCh38
5924836 c.55-4C>T p.? Chr.1: 241548825-241548825 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs189592033 Chr.1: 241548746-241548746 on Build GRCh38
rs751741261 Chr.1: 241548836-241548836 on Build GRCh38
rs758728879 Chr.1: 241548829-241548829 on Build GRCh38
rs779704882 Chr.1: 241548784-241548784 on Build GRCh38
rs1050567035 Chr.1: 241548807-241548807 on Build GRCh38
rs764955806 Chr.1: 241548812-241548812 on Build GRCh38
rs773495621 Chr.1: 241548783-241548783 on Build GRCh38
rs748604072 Chr.1: 241548861-241548861 on Build GRCh38
rs753717195 Chr.1: 241548810-241548810 on Build GRCh38
rs898606851 Chr.1: 241548679-241548679 on Build GRCh38
rs911962259 Chr.1: 241548832-241548832 on Build GRCh38
rs202086485 Chr.1: 241548819-241548819 on Build GRCh38
rs946990907 Chr.1: 241548864-241548864 on Build GRCh38
rs75398491 Chr.1: 241548722-241548722 on Build GRCh38
rs11556464 Chr.1: 241548834-241548834 on Build GRCh38
rs761743645 Chr.1: 241548811-241548811 on Build GRCh38
rs143977547 Chr.1: 241548810-241548811 on Build GRCh38
rs377396383 Chr.1: 241548804-241548804 on Build GRCh38
rs778131618 Chr.1: 241548875-241548875 on Build GRCh38
rs149506288 Chr.1: 241548714-241548714 on Build GRCh38
rs768640176 Chr.1: 241548822-241548822 on Build GRCh38
rs908478914 Chr.1: 241548715-241548715 on Build GRCh38
rs892439828 Chr.1: 241548857-241548857 on Build GRCh38
rs181875734 Chr.1: 241548798-241548798 on Build GRCh38
rs1001961259 Chr.1: 241548667-241548667 on Build GRCh38
rs756582225 Chr.1: 241548872-241548872 on Build GRCh38
rs546081305 Chr.1: 241548692-241548692 on Build GRCh38
rs563055864 Chr.1: 241548696-241548696 on Build GRCh38
rs368539566 Chr.1: 241548846-241548846 on Build GRCh38
rs28431981 Chr.1: 241548862-241548862 on Build GRCh38
rs779754442 Chr.1: 241548811-241548811 on Build GRCh38
rs770046385 Chr.1: 241548781-241548781 on Build GRCh38
rs374808818 Chr.1: 241548724-241548724 on Build GRCh38
rs1032044378 Chr.1: 241548742-241548742 on Build GRCh38
rs896356242 Chr.1: 241548699-241548699 on Build GRCh38
rs771663688 Chr.1: 241548800-241548800 on Build GRCh38

View Full Product Details