Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89726491-89726756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNCA
Entrez Gene ID
6622
Gene Name
synuclein alpha
Gene Aliases
NACP, PARK1, PARK4, PD1
UniGene
Hs.21374
Gene Chromosome Location
Chr.4: 89724099-89838324 on Build GRCh38
Transcripts
NM_000345.3 NM_001146054.1 NM_001146055.1 NM_007308.2 XM_011532208.2 XM_017008562.1 XM_017008563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461644 c.400C>A p.Q134K Chr.4: 89726651-89726651 on Build GRCh38
2957802 c.405C>A p.D135E Chr.4: 89726646-89726646 on Build GRCh38
6469769 c.408C>T p.Y136Y Chr.4: 89726643-89726643 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754479691 Chr.4: 89726590-89726590 on Build GRCh38
rs547270894 Chr.4: 89726717-89726717 on Build GRCh38
rs768678720 Chr.4: 89726675-89726675 on Build GRCh38
rs778588756 Chr.4: 89726605-89726605 on Build GRCh38
rs1030477649 Chr.4: 89726523-89726523 on Build GRCh38
rs955311825 Chr.4: 89726532-89726532 on Build GRCh38
rs144511886 Chr.4: 89726512-89726512 on Build GRCh38
rs370319708 Chr.4: 89726545-89726545 on Build GRCh38
rs199993165 Chr.4: 89726690-89726690 on Build GRCh38
rs1043452934 Chr.4: 89726520-89726520 on Build GRCh38
rs962391035 Chr.4: 89726565-89726565 on Build GRCh38
rs779277002 Chr.4: 89726584-89726584 on Build GRCh38
rs748631342 Chr.4: 89726516-89726516 on Build GRCh38
rs377006107 Chr.4: 89726653-89726653 on Build GRCh38
rs375682339 Chr.4: 89726639-89726639 on Build GRCh38
rs556087044 Chr.4: 89726691-89726691 on Build GRCh38
rs6842093 Chr.4: 89726714-89726714 on Build GRCh38
rs918531737 Chr.4: 89726642-89726642 on Build GRCh38
rs373801428 Chr.4: 89726547-89726547 on Build GRCh38
rs76642636 Chr.4: 89726643-89726643 on Build GRCh38
rs896440399 Chr.4: 89726563-89726563 on Build GRCh38
rs777317657 Chr.4: 89726695-89726696 on Build GRCh38
rs746232417 Chr.4: 89726638-89726638 on Build GRCh38
rs371014092 Chr.4: 89726676-89726676 on Build GRCh38
rs766286972 Chr.4: 89726709-89726709 on Build GRCh38
rs973701461 Chr.4: 89726586-89726586 on Build GRCh38
rs1006605158 Chr.4: 89726544-89726544 on Build GRCh38
rs962490808 Chr.4: 89726712-89726712 on Build GRCh38
rs145304567 Chr.4: 89726551-89726551 on Build GRCh38
rs1052419447 Chr.4: 89726716-89726716 on Build GRCh38
rs749526106 Chr.4: 89726657-89726657 on Build GRCh38
rs756037393 Chr.4: 89726669-89726670 on Build GRCh38
rs781595227 Chr.4: 89726632-89726632 on Build GRCh38
rs773087512 Chr.4: 89726703-89726703 on Build GRCh38
rs757748949 Chr.4: 89726631-89726631 on Build GRCh38
rs778443752 Chr.4: 89726572-89726572 on Build GRCh38
rs755913681 Chr.4: 89726578-89726578 on Build GRCh38
rs755608841 Chr.4: 89726585-89726585 on Build GRCh38
rs760449598 Chr.4: 89726706-89726706 on Build GRCh38
rs747474608 Chr.4: 89726609-89726609 on Build GRCh38
rs71609575 Chr.4: 89726697-89726697 on Build GRCh38
rs772716317 Chr.4: 89726517-89726517 on Build GRCh38
rs372445593 Chr.4: 89726723-89726723 on Build GRCh38
rs753544938 Chr.4: 89726588-89726588 on Build GRCh38
rs537728606 Chr.4: 89726687-89726687 on Build GRCh38

View Full Product Details