Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30580560-30580797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PES1
Entrez Gene ID
23481
Gene Name
pescadillo ribosomal biogenesis factor 1
Gene Aliases
PES
UniGene
Hs.517543
Gene Chromosome Location
Chr.22: 30576625-30607013 on Build GRCh38
Transcripts
NM_001243225.1 NM_001282327.1 NM_001282328.1 NM_014303.3 XM_006724173.1 XM_017028678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1220202 c.1006C>T p.R336* Chr.22: 30580608-30580608 on Build GRCh38
3553365 c.1017C>T p.P339P Chr.22: 30580597-30580597 on Build GRCh38
1261952 c.1043+3_1043+4insTA p.? Chr.22: 30580567-30580568 on Build GRCh38
5941097 c.913-8C>T p.? Chr.22: 30580709-30580709 on Build GRCh38
121529 c.984G>A p.E328E Chr.22: 30580630-30580630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017081841 Chr.22: 30580713-30580713 on Build GRCh38
rs774069893 Chr.22: 30580732-30580732 on Build GRCh38
rs375914441 Chr.22: 30580730-30580730 on Build GRCh38
rs192368103 Chr.22: 30580673-30580673 on Build GRCh38
rs749465412 Chr.22: 30580754-30580754 on Build GRCh38
rs369510488 Chr.22: 30580690-30580690 on Build GRCh38
rs755484176 Chr.22: 30580687-30580687 on Build GRCh38
rs780559156 Chr.22: 30580680-30580680 on Build GRCh38
rs747151927 Chr.22: 30580752-30580752 on Build GRCh38
rs137921166 Chr.22: 30580657-30580657 on Build GRCh38
rs754187400 Chr.22: 30580736-30580736 on Build GRCh38
rs771211816 Chr.22: 30580718-30580718 on Build GRCh38
rs201121515 Chr.22: 30580750-30580750 on Build GRCh38
rs758431732 Chr.22: 30580715-30580715 on Build GRCh38
rs768059240 Chr.22: 30580597-30580597 on Build GRCh38
rs759466296 Chr.22: 30580658-30580658 on Build GRCh38
rs765219602 Chr.22: 30580664-30580664 on Build GRCh38
rs769350869 Chr.22: 30580603-30580603 on Build GRCh38
rs373644713 Chr.22: 30580749-30580749 on Build GRCh38
rs779035902 Chr.22: 30580607-30580607 on Build GRCh38
rs775325322 Chr.22: 30580729-30580729 on Build GRCh38
rs144569778 Chr.22: 30580686-30580686 on Build GRCh38
rs367672411 Chr.22: 30580643-30580643 on Build GRCh38
rs998029705 Chr.22: 30580662-30580662 on Build GRCh38
rs777712062 Chr.22: 30580751-30580751 on Build GRCh38
rs76588799 Chr.22: 30580717-30580717 on Build GRCh38
rs772269763 Chr.22: 30580624-30580624 on Build GRCh38
rs967966887 Chr.22: 30580771-30580771 on Build GRCh38
rs761542093 Chr.22: 30580733-30580733 on Build GRCh38
rs751145692 Chr.22: 30580676-30580676 on Build GRCh38
rs748187531 Chr.22: 30580608-30580608 on Build GRCh38
rs756177566 Chr.22: 30580725-30580725 on Build GRCh38
rs146114623 Chr.22: 30580585-30580585 on Build GRCh38
rs916123360 Chr.22: 30580742-30580742 on Build GRCh38
rs978983552 Chr.22: 30580776-30580776 on Build GRCh38
rs752197414 Chr.22: 30580670-30580670 on Build GRCh38
rs373595484 Chr.22: 30580596-30580596 on Build GRCh38
rs200292450 Chr.22: 30580735-30580735 on Build GRCh38
rs368462198 Chr.22: 30580720-30580720 on Build GRCh38
rs755424997 Chr.22: 30580743-30580743 on Build GRCh38
rs763766110 Chr.22: 30580675-30580675 on Build GRCh38
rs753123216 Chr.22: 30580747-30580747 on Build GRCh38
rs750735932 Chr.22: 30580595-30580595 on Build GRCh38
rs762144194 Chr.22: 30580651-30580653 on Build GRCh38
rs149639037 Chr.22: 30580691-30580691 on Build GRCh38
rs765725755 Chr.22: 30580746-30580746 on Build GRCh38
rs140816492 Chr.22: 30580677-30580677 on Build GRCh38
rs534415458 Chr.22: 30580716-30580716 on Build GRCh38
rs185659178 Chr.22: 30580726-30580726 on Build GRCh38
rs760414430 Chr.22: 30580649-30580649 on Build GRCh38

View Full Product Details