Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57025274-57025545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
UniGene
Hs.528836
Gene Chromosome Location
Chr.16: 56989547-57083524 on Build GRCh38
Transcripts
NM_032206.4 XM_005256193.2 XM_005256194.3 XM_005256195.2 XM_005256196.2 XM_005256197.2 XM_005256199.3 XM_005256201.3 XM_006721297.2 XM_006721298.2 XM_006721300.2 XM_011523373.2 XM_011523375.2 XM_017023766.1 XM_017023767.1 XM_017023768.1 XM_017023769.1 XM_017023770.1 XM_017023771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1217430 c.556G>A p.A186T Chr.16: 57025499-57025499 on Build GRCh38
4061373 c.593A>G p.D198G Chr.16: 57025536-57025536 on Build GRCh38
5595930 c.512C>T p.A171V Chr.16: 57025455-57025455 on Build GRCh38
4061372 c.545G>A p.R182H Chr.16: 57025488-57025488 on Build GRCh38
3754948 c.572C>T p.P191L Chr.16: 57025515-57025515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113208427 Chr.16: 57025368-57025368 on Build GRCh38
rs113392322 Chr.16: 57025296-57025296 on Build GRCh38
rs747884831 Chr.16: 57025298-57025298 on Build GRCh38
rs148647729 Chr.16: 57025406-57025406 on Build GRCh38
rs780366196 Chr.16: 57025493-57025493 on Build GRCh38
rs771246348 Chr.16: 57025323-57025323 on Build GRCh38
rs772254831 Chr.16: 57025449-57025449 on Build GRCh38
rs756533119 Chr.16: 57025327-57025328 on Build GRCh38
rs751376898 Chr.16: 57025399-57025399 on Build GRCh38
rs776988378 Chr.16: 57025479-57025479 on Build GRCh38
rs200019366 Chr.16: 57025407-57025407 on Build GRCh38
rs56132500 Chr.16: 57025329-57025329 on Build GRCh38
rs767160749 Chr.16: 57025488-57025488 on Build GRCh38
rs774696641 Chr.16: 57025333-57025333 on Build GRCh38
rs753388691 Chr.16: 57025351-57025351 on Build GRCh38
rs530594443 Chr.16: 57025393-57025393 on Build GRCh38
rs756373647 Chr.16: 57025421-57025421 on Build GRCh38
rs750291867 Chr.16: 57025490-57025490 on Build GRCh38
rs577082429 Chr.16: 57025299-57025299 on Build GRCh38
rs756288225 Chr.16: 57025491-57025491 on Build GRCh38
rs564063226 Chr.16: 57025453-57025453 on Build GRCh38
rs150502987 Chr.16: 57025306-57025306 on Build GRCh38
rs200485271 Chr.16: 57025466-57025466 on Build GRCh38
rs201860459 Chr.16: 57025372-57025372 on Build GRCh38
rs757451498 Chr.16: 57025421-57025421 on Build GRCh38
rs772690279 Chr.16: 57025510-57025510 on Build GRCh38
rs201453343 Chr.16: 57025460-57025460 on Build GRCh38
rs752310989 Chr.16: 57025378-57025378 on Build GRCh38
rs769212399 Chr.16: 57025332-57025332 on Build GRCh38
rs756392031 Chr.16: 57025353-57025358 on Build GRCh38
rs74439742 Chr.16: 57025515-57025515 on Build GRCh38
rs746124102 Chr.16: 57025328-57025328 on Build GRCh38
rs747438810 Chr.16: 57025516-57025516 on Build GRCh38
rs1003049088 Chr.16: 57025304-57025304 on Build GRCh38
rs769124314 Chr.16: 57025438-57025438 on Build GRCh38
rs774890837 Chr.16: 57025443-57025443 on Build GRCh38
rs778963589 Chr.16: 57025503-57025503 on Build GRCh38
rs762600512 Chr.16: 57025476-57025476 on Build GRCh38
rs748681284 Chr.16: 57025448-57025448 on Build GRCh38
rs769114538 Chr.16: 57025450-57025450 on Build GRCh38
rs139650302 Chr.16: 57025402-57025402 on Build GRCh38
rs765005713 Chr.16: 57025456-57025456 on Build GRCh38
rs781318886 Chr.16: 57025327-57025327 on Build GRCh38
rs954755620 Chr.16: 57025397-57025397 on Build GRCh38
rs766037429 Chr.16: 57025350-57025350 on Build GRCh38
rs375058911 Chr.16: 57025524-57025524 on Build GRCh38
rs763395653 Chr.16: 57025331-57025331 on Build GRCh38
rs749502307 Chr.16: 57025419-57025420 on Build GRCh38
rs75068463 Chr.16: 57025324-57025324 on Build GRCh38
rs779130177 Chr.16: 57025471-57025471 on Build GRCh38
rs749440078 Chr.16: 57025437-57025437 on Build GRCh38
rs371338597 Chr.16: 57025454-57025454 on Build GRCh38
rs1031725966 Chr.16: 57025388-57025388 on Build GRCh38
rs779987156 Chr.16: 57025425-57025425 on Build GRCh38
rs746865619 Chr.16: 57025321-57025321 on Build GRCh38
rs143947320 Chr.16: 57025398-57025398 on Build GRCh38
rs367809019 Chr.16: 57025419-57025419 on Build GRCh38
rs199512041 Chr.16: 57025487-57025487 on Build GRCh38
rs775645807 Chr.16: 57025330-57025330 on Build GRCh38
rs777807836 Chr.16: 57025360-57025361 on Build GRCh38
rs748593399 Chr.16: 57025504-57025504 on Build GRCh38
rs769307574 Chr.16: 57025523-57025523 on Build GRCh38
rs762209630 Chr.16: 57025342-57025342 on Build GRCh38
rs201339650 Chr.16: 57025325-57025325 on Build GRCh38

View Full Product Details