Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 96721790-96722063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTBP2
Entrez Gene ID
58155
Gene Name
polypyrimidine tract binding protein 2
Gene Aliases
PTBLP, brPTB, nPTB
UniGene
Hs.596061
Gene Chromosome Location
Chr.1: 96721605-96815049 on Build GRCh38
Transcripts
NM_001300985.1 NM_001300986.1 NM_001300987.1 NM_001300988.1 NM_001300989.1 NM_001300990.1 NM_021190.3 NR_125357.1 XM_011541874.2 XM_011541875.2 XM_011541876.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6758050 c.6C>T p.D2D Chr.1: 96721870-96721870 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776156545 Chr.1: 96721818-96721818 on Build GRCh38
rs968566009 Chr.1: 96721828-96721828 on Build GRCh38
rs975678448 Chr.1: 96721938-96721938 on Build GRCh38
rs776536700 Chr.1: 96721917-96721917 on Build GRCh38
rs775226037 Chr.1: 96721833-96721833 on Build GRCh38
rs1000020869 Chr.1: 96722035-96722035 on Build GRCh38
rs770422808 Chr.1: 96721816-96721816 on Build GRCh38
rs1030968276 Chr.1: 96722038-96722038 on Build GRCh38
rs935374434 Chr.1: 96721974-96721974 on Build GRCh38
rs1041602146 Chr.1: 96721966-96721966 on Build GRCh38
rs999574097 Chr.1: 96721838-96721838 on Build GRCh38
rs540439062 Chr.1: 96721843-96721843 on Build GRCh38
rs886965953 Chr.1: 96721953-96721953 on Build GRCh38
rs996192317 Chr.1: 96721822-96721822 on Build GRCh38
rs896572698 Chr.1: 96721848-96721848 on Build GRCh38
rs200036230 Chr.1: 96721921-96721921 on Build GRCh38
rs767579562 Chr.1: 96721836-96721836 on Build GRCh38
rs374529117 Chr.1: 96721886-96721886 on Build GRCh38
rs199656587 Chr.1: 96721839-96721839 on Build GRCh38
rs947307251 Chr.1: 96721941-96721941 on Build GRCh38
rs752943417 Chr.1: 96721890-96721890 on Build GRCh38
rs756952411 Chr.1: 96721915-96721915 on Build GRCh38
rs900449436 Chr.1: 96721973-96721973 on Build GRCh38
rs903934423 Chr.1: 96722031-96722031 on Build GRCh38
rs944609118 Chr.1: 96721935-96721935 on Build GRCh38
rs995951477 Chr.1: 96722002-96722002 on Build GRCh38
rs912346169 Chr.1: 96721930-96721930 on Build GRCh38
rs563013908 Chr.1: 96721948-96721948 on Build GRCh38
rs963306477 Chr.1: 96721860-96721860 on Build GRCh38
rs766539552 Chr.1: 96721844-96721844 on Build GRCh38
rs558009316 Chr.1: 96721823-96721823 on Build GRCh38
rs370272686 Chr.1: 96721853-96721853 on Build GRCh38
rs778064495 Chr.1: 96721906-96721906 on Build GRCh38
rs200710034 Chr.1: 96721905-96721905 on Build GRCh38
rs896598725 Chr.1: 96722025-96722025 on Build GRCh38
rs370577082 Chr.1: 96721826-96721826 on Build GRCh38
rs746639232 Chr.1: 96721911-96721911 on Build GRCh38
rs374333701 Chr.1: 96721922-96721922 on Build GRCh38
rs746695158 Chr.1: 96721814-96721814 on Build GRCh38
rs1056383926 Chr.1: 96722018-96722018 on Build GRCh38
rs1043573465 Chr.1: 96722028-96722028 on Build GRCh38
rs992418966 Chr.1: 96721852-96721852 on Build GRCh38
rs761908551 Chr.1: 96721934-96721934 on Build GRCh38
rs375724475 Chr.1: 96721851-96721851 on Build GRCh38
rs752796391 Chr.1: 96721889-96721890 on Build GRCh38
rs773472817 Chr.1: 96721837-96721837 on Build GRCh38
rs948221846 Chr.1: 96721857-96721857 on Build GRCh38
rs1040285303 Chr.1: 96721946-96721946 on Build GRCh38
rs974264039 Chr.1: 96721904-96721904 on Build GRCh38
rs201636537 Chr.1: 96721859-96721859 on Build GRCh38
rs978109281 Chr.1: 96721830-96721830 on Build GRCh38
rs762812836 Chr.1: 96721835-96721835 on Build GRCh38
rs950987148 Chr.1: 96721910-96721910 on Build GRCh38

View Full Product Details