Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 57005603-57005838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2B
Entrez Gene ID
5431
Gene Name
RNA polymerase II subunit B
Gene Aliases
POL2RB, RPB2, hRPB140
UniGene
Hs.602757
Gene Chromosome Location
Chr.4: 56978640-57031162 on Build GRCh38
Transcripts
NM_000938.2 NM_001303268.1 NM_001303269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
402449 c.1217+1G>T p.? Chr.4: 57005720-57005720 on Build GRCh38
7418178 c.1150G>T p.D384Y Chr.4: 57005652-57005652 on Build GRCh38
3302325 c.1194G>A p.P398P Chr.4: 57005696-57005696 on Build GRCh38
6340120 c.1217G>T p.G406V Chr.4: 57005719-57005719 on Build GRCh38
4895752 c.1206C>T p.F402F Chr.4: 57005708-57005708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528122351 Chr.4: 57005774-57005774 on Build GRCh38
rs755244443 Chr.4: 57005663-57005663 on Build GRCh38
rs770507784 Chr.4: 57005706-57005706 on Build GRCh38
rs375305377 Chr.4: 57005722-57005722 on Build GRCh38
rs866943329 Chr.4: 57005673-57005674 on Build GRCh38
rs779239593 Chr.4: 57005672-57005672 on Build GRCh38
rs369396992 Chr.4: 57005743-57005743 on Build GRCh38
rs758840553 Chr.4: 57005693-57005693 on Build GRCh38
rs751881138 Chr.4: 57005751-57005751 on Build GRCh38
rs954968231 Chr.4: 57005749-57005749 on Build GRCh38
rs199642101 Chr.4: 57005695-57005695 on Build GRCh38
rs754026412 Chr.4: 57005642-57005642 on Build GRCh38
rs564863397 Chr.4: 57005807-57005807 on Build GRCh38
rs962064271 Chr.4: 57005776-57005776 on Build GRCh38
rs775358584 Chr.4: 57005755-57005755 on Build GRCh38
rs371510241 Chr.4: 57005735-57005735 on Build GRCh38
rs748583124 Chr.4: 57005682-57005682 on Build GRCh38
rs139369251 Chr.4: 57005779-57005779 on Build GRCh38
rs746456044 Chr.4: 57005696-57005696 on Build GRCh38

View Full Product Details